Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 10
Immunogen sequence: IKAGSRYEDF SNLGTTHLLR LTSSLTTKGA SSFKITRGIE AVGGKLSVTA TRENMAYTVE CLRGDVDILM EFLLNVTTAP EFRRWEVADL QPQLKIDKAV AFQNPQTHVI ENLHAAAYRN ALANPLYCPD YRIGKVTSEE LHYFVQN
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 2; Core protein II; Cytochrome b-c1 complex subunit 2, mitochondrial; cytochrome bc-1 complex core protein II; U MC3DN5; Ubiquinol-cytochrome-c reductase complex core protein 2
Gene Aliases: MC3DN5; QCR2; UQCR2; UQCRC2
UniProt ID: (Human) P22695
Entrez Gene ID: (Human) 7385
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support