Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: KANYRGGEIR EQNGDSLVHA AFVAESAVAG SAEANAFSVL QHVLGAGPHV KRGSNTTSHL HQAVAKATQQ PFDVSAFNAS YSDSGLFGIY TISQATAAGD V
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 2; Core protein II; Cytochrome b-c1 complex subunit 2, mitochondrial; cytochrome bc-1 complex core protein II; U MC3DN5; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; Ubiquinol-cytochrome-c reductase complex core protein 2
Gene Aliases: 1500004O06Rik; MC3DN5; QCR2; UQCR2; UQCRC2
UniProt ID: (Human) P22695, (Mouse) Q9DB77, (Rat) P32551
Entrez Gene ID: (Human) 7385, (Mouse) 67003, (Rat) 293448
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support