Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 8; complex III subunit VII; Complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); ubiquinol-cytochrome c reductase binding protein; Ubiquinol-cytochrome c reductase complex 9.5 kDa protein; Ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa
Gene Aliases: 1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5kDa; AA959903; c1502; MC3DN4; QCR8; QP-C; QPC; UQCR7; Uqcrb; UQCRQ
UniProt ID: (Human) O14949, (Mouse) Q9CQ69
Entrez Gene ID: (Human) 27089, (Mouse) 22272
Molecular Function: oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support