Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEANGLGPQ GFPELKNDTF LRAAWGEETD YTPVWCMRQA GRYLPEFRET RAAQDFFSTC RSPEACCELT LQPLRRFPLD AAIIFSDILV VPQALGMEVT MVPGKGPSFP EPLREEQDLE RLRDPEVVAS ELGYVFQAIT LTRQRLAGRV PLIGFAGAPW TLMTYMVEGG GSSTMAQAKR WLYQRPQASH QLLRILTDAL VPYLVGQVVA GAQALQLFES HAGHLGPQLF NKFALPYIRD VAKQVKARLR EAGLAPVPMI IFAKDGHFAL EELAQAGYEV VGLDWTVAPK KARECVGKTV TLQVNLDPCA LYASEEEIGQ LVKQMLDDFG PHRYIANLGH GLYPDMDPEH VGAFVDAVHK HSRLLRQN (1-367 aa encoded by BC001778)
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: porphyrinogen carboxylase; UPD; URO D; URO-D; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase
Gene Aliases: AI323803; PCT; porphyrinogen carboxy-lyase; UPD; Uro-d; UROD
UniProt ID: (Human) P06132, (Mouse) P70697
Entrez Gene ID: (Human) 7389, (Mouse) 22275, (Rat) 29421
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support