Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ALEELAQAGY EVVGLDWTVA PKKARECVGK TVTLQGNLDP CALYASEEEI GQLVKQMLDD FGPHRYIANL GHGLYPDMDP EHVGAFVDAV HKHSRLLRQN
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPD; URO D; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase
Gene Aliases: PCT; UPD; UROD
UniProt ID: (Human) P06132
Entrez Gene ID: (Human) 7389
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support