Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MKGQQKTAET EEGTVQIQEG AVATGEDPTS VAIASIQSAA TFPDPNVKYV FRTENGGQVM YRVIQVSEGQ LDGQTEGTGA ISGYPATQSM TQAVIQGAFT SDDAVDTEGT AAETHYTYFP STAVGDGAGG TTSGSTAAVV TTQGSEALLG QATPPGTGQF FVMMSPQEVL QGGSQRSIAP RTHPYSPKSE APRTTRDEKR RAQHNEVERR RRDKINNWIV QLSKIIPDCS MESTKSGQSK GGILSKACDY IQELRQSNHR LSEELQGLDQ LQLDNDVLRQ QVEDLKNKNL LLRAQLRHHG LEVVIKNDSN
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb11; Class B basic helix-loop-helix protein 11; Major late transcription factor 1; Upstream stimulatory factor 1; usf1-bd
Gene Aliases: BHLHB11; FCHL; FCHL1; HYPLIP1; MLTF; MLTFI; UEF; USF; USF1
UniProt ID: (Human) P22415
Entrez Gene ID: (Human) 7391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support