Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGPESDAQ YQFTGIKKYF NSYTLTGRMN CVLATYGSIA LIVLYFKLRS KKTPAVKAT (1-58 aa encoded by BC072683)
Plays a critical role in maintaining the ATP synthase population in mitochondria. Target protein is a novel protein that is upregulated in streptozotocin-induced diabetic rats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase membrane subunit DAPIT, mitochondrial; ATP synthase membrane subunit K, mitochondrial; Diabetes Associated Protein in Insulin-sensitive Tissues; Diabetes-associated protein in insulin-sensitive tissues; HCV F-transactivated protein 2; Up-regulated during skeletal muscle growth protein 5; upregulated during skeletal muscle growth 5; upregulated during skeletal muscle growth 5 homolog
Gene Aliases: 2010301L15Rik; ATP5MD; ATP5MK; bA792D24.4; DAPIT; HCVFTP2; PD04912; USMG5
UniProt ID: (Human) Q96IX5, (Rat) Q9JJW3, (Mouse) Q78IK2
Entrez Gene ID: (Human) 84833, (Rat) 171069, (Mouse) 66477
Molecular Function: ATP synthase primary active transporter transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support