Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Deubiquitinating enzyme. Has almost no deubiquitinating activity by itself and requires the interaction with WDR20 and WDR48 to have a high activity (PubMed:19075014, PubMed:27373336). Not involved in deubiquitination of monoubiquitinated FANCD2 (PubMed:19075014). In complex with WDR48, acts as a potential tumor suppressor by positively regulating PHLPP1 stability (PubMed:24145035). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 12; Ubiquitin carboxyl-terminal hydrolase 12; Ubiquitin specific peptidase 12; ubiquitin specific protease 12 like 1; Ubiquitin thioesterase 12; ubiquitin thiolesterase 12; Ubiquitin-hydrolyzing enzyme 1; Ubiquitin-specific-processing protease 12
Gene Aliases: UBH1; USP12; USP12L1
UniProt ID: (Human) O75317
Entrez Gene ID: (Human) 219333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support