Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
Immunogen sequence: MAEGGAADL DTQRSDIATL LKTSLRKGDT WYLVDSRWFK QWKKYVGFDS WDKYQMGDQN VYPGPIDNSG LLKDGDAQSL KEHLIDELDY ILLPTEGWNK LVSWYTLMEG QEPIARKVVE QGMFVKHCKV EVYLTELKLC ENGNMNNVVT RRFSKADTID TIEKEIRKIF SIPDEKETRL WNKYMSNTFE PLNKPDSTIQ DAGLYQGQVL VIEQKNEDGT WPRGPSTPKK PLEQSC (1-235 aa encoded by B C020688)
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 15; deubiquitinating enzyme Ubp109; granule cell antiserum positive 18; MGC131982; MGC149838; MGC74854; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin carboxyl-terminal hydrolase of 109 kDa; ubiquitin specific protease 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; Unph4
Gene Aliases: 4921514G19Rik; AI327321; E430033I05Rik; Gcap18; KIAA0529; Ubp109; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y4E8, (Mouse) Q8R5H1, (Rat) Q9R085
Entrez Gene ID: (Human) 9958, (Mouse) 14479, (Rat) 171329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support