Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGKKRTKGK TVPIDDSSET LEPVCRHIRK GLEQGNLKKA LVNVEWNICQ DCKTDNKVKD KAEEETEEKP SVWLCLKCGH QGCGRNSQEQ HALKHYLTPR SEPHCLVLSL DNWSVWCYVC DNEVQYCSSN QLGQVVDYVR KHASITTPKP EKDNGNIELE NKKLEKESKN EQEREKKENM AKENPPMNSP CQITVKGLSN LGNTCFFNAV MQNLSQTPVL RELLKEVKMS GTIVKIEPPD LALTEPLEIN LEPPGPLTLA MSQFLNEMQE TKKGVVTPKE LFSQVCKKAV RFKGYQQQDS QELLRYLLDG MRAEEHQRVS KGILKAFGNS TEKLDEELKN KVKDYEKKKS MPSFVDRIFG GELTSMIMCD QCRTVSLVHE SFLDLSL (1-386 aa encoded by BC030777)
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 16; deubiquitinating enzyme 16 {ECO:0000255|HAMAP-Rule:MF_03062}; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255|HAMAP-Rule:MF_03062}; ubiquitin specific protease 16; Ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255|HAMAP-Rule:MF_03062}; ubiquitin thiolesterase 16; Ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; Ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255|HAMAP-Rule:MF_03062}; USP16
Gene Aliases: 1200004E02Rik; 2810483I07Rik; 6330514E22Rik; MSTP039; UBP-M; UBPM; USP16
UniProt ID: (Human) Q9Y5T5, (Rat) Q2KJ09, (Mouse) Q99LG0
Entrez Gene ID: (Human) 10600, (Rat) 288306, (Mouse) 74112
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support