Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSGGASATGP RRGPPGLEDT TSKKKQKDRA NQESKDGDPR KETGSRYVAQ AGLEPLASGD PSASASHAAG ITGSRHRTRL FFPSSSGSAS TPQEEQTKEG ACEDPHDLLA TPTPELLLDW RQSAEEVIVK LRVGVGPLQL EDVDAAFTDT DCVVRFAGGQ QWGGVFYAEI KSSCAKVQTR KGSLLHLTLP KKVPMLTWPS LLVEADEQLC IPPLNSQTCL LGSEENLAPL AGEKAVPPGN DPVSPAMVRS RNPGKDDCAK EEMAVAADAA TLVDEPESMV NLAFVKNDSY EKGPDSVVVH VYVKEICRDT SRVLFREQDF TLIFQTRDGN FLRLHPGCGP HTTFRWQVKL RNLIEPEQCT FCFTASRIDI CLRKRQSQRW GGLEAPAARV GGAKVAVPTG PTPLDSTPPG GAPHPLTGQE EARAVEKDKS KARSEDTGLD SVATRTPMEH VTPKPETHLA SPKPTCMVPP MPHSPVSGDS VEEEEEEEKK VCLPGFTGLV NLGNTCFMNS VIQSLSNTRE LRDFFHDRSF EAEINYNNPL GTGGRLAIGF AVLLRALWKG THHAFQPSKL KAIVASKASQ FTGYAQHDAQ EFMAFLLDGL HEDLNRIQNK PYTETVDSDG RPDEVVAEEA WQRHKMRNDS FIVDLFQGQY KSKLVCPVCA KVSITFDPFL YLPVPLPQKQ KVLPVFYFAR EPHSKPIKFL VSVSKENSTA SEVLDSLSQS VHVKPENLRL AEVIKNRFHR VFLPSHSLDT VSPSDTLLCF ELLSSELAKE RVVVLEVQQR PQVPSVPISK CAACQRKQQS EDEKLKRCTR CYRVGYCNQL CQKTHWPDHK GLCRPENIGY PFLVSVPASR LTYARLAQLL EGYARYSVSV FQPPFQPGRM ALESQSPGCT TLLSTGSLEA GDSERDPIQP PELQLVTPMA EGDTGLPRVW AAPDRGPVPS TSGISSEMLA SGPIEVGSLP AGERVSRPEA AVPGYQHPSE AMNAHTPQFF IYKIDSSNRE QRLEDKGDTP LELGDDCSLA LVWRNNERLQ EFVLVASKEL ECAEDPGSAG EAARAGHFTL DQCLNLFTRP EVLAPEEAWY CPQCKQHREA SKQLLLWRLP NVLIVQLKRF SFRSFIWRDK INDLVEFPVR NLDLSKFCIG QKEEQLPSYD LYAVINHYGG MIGGHYTACA RLPNDRSSQR SDVGWRLFDD STVTTVDESQ VVTRYAYVLF YRRRNSPVER PPRAGHSEHH PDLGPAAEAA ASQASRIWQE LEAEEEPVPE GSGPLGPWGP QDWVGPLPRG PTTPDEGCLR YFVLGTVAAL VALVLNVFYP LVSQSRWR
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs), 1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 19; Ubiquitin carboxyl-terminal hydrolase 19; ubiquitin specific protease 19; Ubiquitin thioesterase 19; ubiquitin thiolesterase 19; Ubiquitin-specific-processing protease 19; Zinc finger MYND domain-containing protein 9
Gene Aliases: KIAA0891; USP19; ZMYND9
UniProt ID: (Human) O94966
Entrez Gene ID: (Human) 10869
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support