Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: MSGGASATGP RRGPPGLEDT TSKKKQKDRA NQESKDGDPR KETGSRYVAQ AGLEPLASGD PSASASHAAG ITGSRHRTRL FFPSSSGSAS TPQEEQTKEG ACEDPHDLLA TPTPELLLDW RQSAEEVIVK LRVGVGPLQL EDVDAAFTDT DCVVRFAGGQ QWGGVFYAEI KSSCAKVQTR KGSLLHLTLP KKVPMLTWPS; Positive Samples: U-87MG, A-431, HeLa; Cellular Location: Endoplasmic reticulum membrane, Single-pass membrane protein
Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs), 1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 19; Ubiquitin carboxyl-terminal hydrolase 19; ubiquitin specific protease 19; Ubiquitin thioesterase 19; ubiquitin thiolesterase 19; ubiquitin-specific processing protease 19; Ubiquitin-specific-processing protease 19; Zinc finger MYND domain-containing protein 9
Gene Aliases: 8430421I07Rik; AI047774; KIAA0891; USP19; ZMYND9
UniProt ID: (Human) O94966, (Mouse) Q3UJD6
Entrez Gene ID: (Human) 10869, (Mouse) 71472
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support