Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • USP20 Antibodies

            Bethyl Laboratories

            USP20/VDU2 Polyclonal Antibody

            View all (9) USP20 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite USP20/VDU2 Polyclonal Antibody

            • Antibody Testing Data (2)
            Application
            USP20/VDU2 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            USP20/VDU2 Antibody (A301-189A) in WB

            Detection of mouse USP20/VDU2 by western blot. Samples: Whole cell lysate (50 µg) from NIH 3T3, TCMK-1, 4T1, CT26.WT, and rat C6 cells. Antibodies: Affinity purified rabbit anti-USP20/VDU2 antibody (Product # A301-189A) (lot A301-189A-2) used for WB at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes.
            View Product

            USP20/VDU2 Antibody in Western Blot (WB)
            USP20/VDU2 Antibody in Western Blot (WB)

            Product Details

            A301-189A

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:2,000-1:10,000
            -

            Immunoprecipitation (IP)

            2-5 µg/mg lysate
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Region between residue 863 and 913 of Human ubiquitin specific peptidase 20 (pVHL-interacting deubiquitinating enzyme 2).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            tris citrate/phosphate, pH 7-8

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            The recommended shelf life for this product is 1 year from date of receipt.

            Target Information

            SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Deubiquitinating enzyme 20; hVDU2; pVHL-interacting deubiquitinating enzyme 2; Ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific protease 20; Ubiquitin thioesterase 20; ubiquitin thiolesterase 20; Ubiquitin-specific-processing protease 20; VHL-interacting deubiquitinating enzyme 2

            View more View less

            Gene Aliases: 1700055M05Rik; AI467231; hVDU2; KIAA1003; LSFR3A; USP20; VDU2

            View more View less

            UniProt ID: (Human) Q9Y2K6, (Mouse) Q8C6M1

            View more View less

            Entrez Gene ID: (Human) 10868, (Mouse) 74270

            View more View less

            Function(s)

            G-protein coupled receptor binding cysteine-type endopeptidase activity thiol-dependent ubiquitin-specific protease activity protein binding zinc ion binding peptidase activity cysteine-type peptidase activity hydrolase activity thiol-dependent ubiquitinyl hydrolase activity metal ion binding cysteine protease

            Process(es)

            ubiquitin-dependent protein catabolic process endocytosis regulation of G-protein coupled receptor protein signaling pathway protein deubiquitination protein K63-linked deubiquitination protein K48-linked deubiquitination proteolysis proteasome-mediated ubiquitin-dependent protein catabolic process regulation of proteasomal protein catabolic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-pxx2l:80/100.66.78.53:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0