Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTGSNSHIT ILTLKVLPHF ESLGKQEKIP NKMSAFRNHC PHLDSVGEIT KEDLIQKSLG TCQDCKVQGP NLWACLENRC SYVGCGESQV DHSTIHSQET KHYLTVNLTT LRVWCYACSK EVFLDRKLGT QPSLPHVRQP HQIQENSVQD FKIPSNTTLK TPLVAVFDDL DIEADEEDEL RARGLTGLKN IGNTCYMNAA LQALSNCPPL TQFFLDCGGL ARTDKKPAIC KSYLKLMTEL WHKSRPGSVV PTTLFQGIKT VNPTFRGYSQ QDAQEFLRCL MDLLHEELKE QVMEVEEDPQ T (1-300 aa encoded by BC016663)
DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 33; hVDU1; pVHL-interacting deubiquitinating enzyme 1; Ubiquitin carboxyl-terminal hydrolase 33; ubiquitin specific protease 33; Ubiquitin thioesterase 33; ubiquitin thiolesterase 33; Ubiquitin-specific-processing protease 33; VHL-interacting deubiquitinating enzyme 1; Vhlh-interacting deubiquitinating enzyme 1
Gene Aliases: 9830169D19Rik; AA409780; KIAA1097; USP33; VDU1
UniProt ID: (Human) Q8TEY7, (Mouse) Q8R5K2
Entrez Gene ID: (Human) 23032, (Mouse) 170822, (Rat) 310960
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support