Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 'Lys-4' trimethylation (H3K4).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 36; Ubiquitin carboxyl-terminal hydrolase 36; ubiquitin specific protease 36; Ubiquitin thioesterase 36; Ubiquitin-specific-processing protease 36
Gene Aliases: DUB1; KIAA1453; USP36
UniProt ID: (Human) Q9P275
Entrez Gene ID: (Human) 57602
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support