Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Mouse liver. The target is usually found in the following locations: Nucleus.
Immunogen sequence: KNPTIVNFPI TNVDLREYLS EEVQAVHKNT TYDLIANIVH DGKPSEGSYR IHVLHHGTGK WYELQDLQVT DILPQMITLS EAYIQIWKRR DNDETNQQGA
USP39 may play a role in mRNA splicing. It is unsure if the protein really exhibits hydrolase activity. It could be a competitor of ubiquitin C-terminal hydrolases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inactive ubiquitin-specific peptidase 39; SAD1 homolog; small nuclear ribonucleoprotein 65kDa (U4/U6.U5); SnRNP assembly defective 1 homolog; U4/U6.U5 tri-snRNP-associated 65 kDa protein; U4/U6.U5 tri-snRNP-associated protein 2; Ubiquitin carboxyl-terminal hydrolase 39; ubiquitin specific protease 39
Gene Aliases: 65K; AA408960; AI894154; CGI-21; D6Wsu157e; HSPC332; PRO2855; SAD1; SNRNP65; USP39
UniProt ID: (Human) Q53GS9, (Mouse) Q3TIX9
Entrez Gene ID: (Human) 10713, (Rat) 297336, (Mouse) 28035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support