Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSLKDLNKKT EVKPEKISTK SYVHSALKIF KTAEECRLDR DEERAYVLYM KYVTVYNLIK KRPDFKQQQD YFHSILGPGN IKKAVEEAER LSESLKLRYE EAEVRKKLEE KDRQEEAQRL QQKRQETGRE DGGTLAKGSL ENVLDSKDKT QKSNGEKNEK CETKEKGAIT AKELYTMMTD KNISLIIMDA RRMQDYQDSC ILHSLSVPEE AISPGVTASW IEAHLPDDSK DTWKKRGNV
USP8 is a ubiquitin specific protease that plays an important regulatory role at the level of protein turnover by preventing degradation. USP8 is involved in cell proliferation, and probably regulates the stability of STAM2 and RASGRF1. USP8 may regulate T-cell energy mediated by RNF128 via the formation of a complex containing RNF128 and STAM2. As revealed by structure/function studies, USP8 forms a ternary complex with RNF128 and OTUB1, and interacts with the SH3 domain of STAM2 and RASGRF1. Expression of USP8 is induced Upon growth stimulation in starved human fibroblasts, and expression decreases in response to growth arrest induced by cell-cell contact.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 8; hUBPy; mUBPy; putative deubiquitinating enzyme; Ubiquitin carboxyl-terminal hydrolase 8; Ubiquitin isopeptidase Y; Ubiquitin thioesterase 8; ubiquitin thiolesterase 8; Ubiquitin-specific-processing protease 8
Gene Aliases: AI574262; AW557536; HumORF8; KIAA0055; mKIAA0055; SPG59; UBPY; USP8
UniProt ID: (Human) P40818, (Mouse) Q80U87
Entrez Gene ID: (Human) 9101, (Mouse) 84092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support