Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLSPTPVSTE LSENGEGDFR YLGMGDSHIP PPVPSEFNDV SQNTHLRQDH NYCSPTKKNP CEVQPDSLTN NACVRTLNLE SPMKTDIFDE FFSSSALNAL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: highly charged protein; SUMO-specific isopeptidase USPL1; Ubiquitin-specific peptidase-like protein 1; USP-like 1
Gene Aliases: bA121O19.1; C13orf22; D13S106; D13S106E; USPL1
UniProt ID: (Human) Q5W0Q7
Entrez Gene ID: (Human) 10208
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support