Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: DHSDSESTSS DNSGRRRKGP FKTIKFGTNI DLSDDKKWKL QLHELTKLPA FVRVVSAGNL LSHVGHTILG MNTVQLYMKV PGSRTPGHQE NNNFCSVNIN IGPGDCEWFV VPEGYWGVLN DFCEKNNLNF LMGSWWPNLE DLYEANVPVY RFIQRPGDLV WINAGTVHWV QAIGWCNNIA WNVGPLTACQ YKLAVERYEW NKLQSVKSIV PMVHLSWNMA RNIKVSDPKL FEMIKYCLLR TLKQCQTLRE ALIAAGKEII WHGRTKEEPA HYCSICEVEV FDLLFVTNES NSRKTYIVHC QDCARKTSGN LENFVVLEQY KMEDLMQVYD QFTLAPPLPS ASS
This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(27) demethylase 6A; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); Histone demethylase UTX; lysine (K)-specific demethylase 6A; Lysine-specific demethylase 6A; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein
Gene Aliases: bA386N14.2; KABUK2; KDM6A; UTX
UniProt ID: (Human) O15550
Entrez Gene ID: (Human) 7403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support