Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSASASVGGQ VPQPPPGPAA ALPPGSAARA LHVELPSQQR RLRHLRNIAA RNIVNRNGHQ LLDTYFTLHL CSTEKIYKEF YRSEVIKNSL NPTWRSLDFG IMPDRLDTSV SCFVVKIWGG KENIYQLLIE WKVCLDGLKY LGQQIHARNQ NEIIFGLNDG YYGAPFEHKG YSNAQKTILL QVDQNCVRNS YDVFSLLRLH RAQCAIKQTQ VTVQKIGKEI EEKLRLTSTS NELKKKSECL QLKILVLQNE LERQKKALGR EVALLHKQQI ALQDKGSAFS AEHLKLQLQK ESLNELRKEC TAKRELFLKT NAQLTIRCRQ LLSELSYIYP IDLNEHKDYF VCGVKLPNSE DFQAKDDGSI AVALGYTAHL VSMISFFLQV PLRYPIIHKG SRSTIKDNIN DKLTEKEREF PLYPKGGEKL QFDYGVYLLN KNIAQLRYQH GLGTPDLRQT LPNLKNFMEH GLMVRCDRHH TSSAIPVPKR QSSIFGGADV GFSGGIPSPD KGHRKRASSE NERLQYKTPP PSYNSALAQP VTTVPSMGET ERKITSLSSS LDTSLDFSKE NKKKGEDLVG SLNGGHANVH PSQEQGEALS GHRATVNGTL LPSEQAGSAS VQLPGEFHPV SEAELCCTVE QAEEIIGLEA TGFASGDQLE AFNCIPVDSA VAVECDEQVL GEFEEFSRRI YALNENVSSF RRPRRSSDK
UVRAG is predicted to contain a coiled-coil structure and is likely to be metabolically unstable based on the occurrence of a strong PEST domain. It interacts with Beclin 1, leading to activation of autophagy and thereof inhibition of tumorigenicity of human colon cancer cells. The gene for UVRAG protein is localized on human chromosome 11q13. 5 and complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells. The gene has been extremely well conserved during evolution, implying an important role for this gene product in cell metabolism. Chromosomal aberrations involving this gene are associated with left-right axis malformation, and mutations in this gene have been associated with colon cancer. The protein is highly expressed in human brain, lung, kidney and liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beclin 1 binding protein; disrupted in heterotaxy; p63; UV radiation resistance-associated gene protein
Gene Aliases: DHTX; p63; UVRAG; VPS38
UniProt ID: (Human) Q9P2Y5
Entrez Gene ID: (Human) 7405
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support