Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
UVRAG is predicted to contain a coiled-coil structure and is likely to be metabolically unstable based on the occurrence of a strong PEST domain. It interacts with Beclin 1, leading to activation of autophagy and thereof inhibition of tumorigenicity of human colon cancer cells. The gene for UVRAG protein is localized on human chromosome 11q13. 5 and complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells. The gene has been extremely well conserved during evolution, implying an important role for this gene product in cell metabolism. Chromosomal aberrations involving this gene are associated with left-right axis malformation, and mutations in this gene have been associated with colon cancer. The protein is highly expressed in human brain, lung, kidney and liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beclin 1 binding protein; disrupted in heterotaxy; p63; UV radiation resistance-associated gene protein; UV radiation resistance-associated protein
Gene Aliases: 9530039D02Rik; AI648770; BB124205; DHTX; p63; UVRAG; Uvrag1; Uvragl; VPS38
UniProt ID: (Human) Q9P2Y5, (Mouse) Q8K245
Entrez Gene ID: (Human) 7405, (Chicken) 419091, (Dog) 607998, (Rat) 308846, (Mouse) 78610
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support