Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
This antibody is predicted to react with Mouse, Rat, Bovine.
Cellular senescence is a hallmark of tumor suppression and tissue aging. Senescent cells contain domains of heterochromatin, called senescence-associated heterochromatin foci (SAHF), that repress proliferation-promoting genes. The protein encoded by this gene binds to proliferation-promoting genes and is required for SAHF formation, enhancing methylation of histone H3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIRA-binding protein; Protein VT4; Ubinuclein-1; Ubiquitously expressed nuclear protein
Gene Aliases: UBN1; VT; VT4
UniProt ID: (Human) Q9NPG3
Entrez Gene ID: (Human) 29855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support