Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody detects 70kD band of UBQLN1.
Immunogen sequence: MAESGESGG PPGSQDSAAG AEGAGAPAAA ASAEPKIMKV TVKTPKEKEE FAVPENSSVQ QFKEEISKRF KSHTDQLVLI FAGKILKDQD TLSQHGIHDG LTVHLVIKTQ NRPQDHSAQQ TNTAGSNVTT SSTPNSNSTS GSATSNPFGL GGLGGLAGLS SLGLNTTNFS ELQSQMQRQL LSNPEMMVQI MENPFVQSML SNPDLMRQLI MANPQMQQLI QRNPEISHML NNPDIMRQTL ELARNPAMMQ EMMRNQDRAL SNLESIPGGY NALRRMYTDI QEPMLSAAQE QFGGNPFASL VSNTSSGEGS QPSRTENRDP LPNPWAPQTS QSSSASSGTA STVGGTTGST ASGTSGQSTT APNLVPGVGA SMFNTPGMQS LLQQITENPQ LMQNMLSAPY MRSMMQSLSQ NPDLAAQMQN PDTLSAMSNP RAMQALLQIQ QGLQTLATEA PGLIPGFTPG LGALGSTGGS SGTNGSNATP SENTSPTAGT TEPGHQQFIQ QMLQALAGVN PQLQNPEVRF QQQLEQLSAM GFLNREANLQ ALIATGGDIN AAIERLLGSQ PS (1-561 aa encoded by BC010066)
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer's and Parkinson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPLIC-1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; Ubiquilin-1
Gene Aliases: DA41; DSK2; PLIC-1; PLIC1; UBQLN1; UBQN; XDRP1
UniProt ID: (Human) Q9H3R4, (Rat) Q9JJP9
Entrez Gene ID: (Human) 29979, (Rat) 114590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support