Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H2A histone family, member R; H2A-clustered histone 1; H2ALys119ub; histone 1, H2aa; Histone H2A type 1-A; Histone H2A type 4; Histone H2A, testis; Histone H2A/r; testis-specific histone 2a; TH2A; Ubiquityl-Histone 2A K119; Ubiquityl-Histone 2A Lys119
Gene Aliases: bA317E16.2; H2AA; H2AC1; H2AFR; HIST1H2AA; TH2A
UniProt ID: (Human) Q96QV6, (Rat) Q00728
Entrez Gene ID: (Human) 221613, (Rat) 24828
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support