Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TQSVYIQYQ GFRVQLESMK KLSDLEAQWA PSPRLQAQSL LPAVCHHPAL PQDLQPVCAS QEASSIFKTL RTIANDDCEL CVNVACTGCL (24-112 aa encoded by BC093779)
Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins. May be a potent physiological regulator of intestinal fluid and electrolyte transport. May be an autocrine/paracrine regulator of intestinal salt and water transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: granule cell antiserum positive 2; Guanylate cyclase activator 2B; guanylate cyclase activator 2b (retina); guanylate cyclase activator 2B (uroguanylin); prepro-uroguanylin; uroguanylin
Gene Aliases: AV066530; GCAP-II; Gcap2; GUCA2B; UGN
UniProt ID: (Human) Q16661, (Mouse) O09051, (Rat) P70668
Entrez Gene ID: (Human) 2981, (Mouse) 14916, (Rat) 64055
Molecular Function:
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support