Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
FIGURE: 1 / 1
Positive Control:Human urinary bladder.
Cellular Location: Cell surface. Heterodimer formation with UPK1B is a prerequisite to exit out of the endoplasmic reticulum.
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ32198; MGC10902; p35; UP3b; UPIIIb; Uroplakin IIIb; Uroplakin-3b
Gene Aliases: UPK3B
UniProt ID: (Human) Q9BT76
Entrez Gene ID: (Human) 105375355
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support