Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
ATP6V1C1 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1; FLJ20057; H(+)-transporting two-sector ATPase, subunit C; H+ -ATPase C subunit; H+-transporting ATPase chain C, vacuolar; subunit C of vacuolar proton-ATPase V1 domain; testicular tissue protein Li 223; V-ATPase C subunit; V-ATPase subunit C 1; V-type proton ATPase subunit C 1; vacuolar ATP synthase subunit C; vacuolar proton pump C subunit; Vacuolar proton pump subunit C 1; vacuolar proton pump, 42-kD subunit; vacuolar proton-ATPase, subunit C, VI domain
Gene Aliases: ATP6C; ATP6D; ATP6V1C1; VATC; Vma5
UniProt ID: (Human) P21283
Entrez Gene ID: (Human) 528
Molecular Function: ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support