Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: human VAMP5-transfected 293T
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myobrevin; vamp; VAMP-5; Vesicle-associated membrane protein 5
Gene Aliases: HSPC191; VAMP5
UniProt ID: (Human) O95183
Entrez Gene ID: (Human) 10791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support