Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDTESTYSGY SYYSSHSKKS HRQGERTRER HKSPRNKDGR GSEKSVTIQP PTGEPLLGND STRTEEVQDD NWGETTTAIT GTSEHSISQE DIARISKDME DSVGLDCKRY LGLTVASFLG LLVFLTPIAF ILLPPILWRD ELEPCGTICE GLFISMAFKL LILLIGTWAL FFRKRRADMP RVFVFRALLL VLIFLFVVSY WLFYGVRILD SRDRNYQGIV QYAVSLVDAL LFIHYLAIVL LELRQLQPMF TLQVVRSTDG ESRFYSLGHL SIQRAALVVL ENYYKDFTIY NPNLLTASKF RAAKHMAGLK VYNVDGPSNN ATGQSRAMIA AAARRRDSSH NELYYEEAEH ERRVKKRKAR LVVAVEEAFI HIQRLQAEEQ QKAPGEVMDP REAAQAIFPS MARALQKYLR ITRQQNYHSM ESILQHLAFC ITNGMTPKAF LERYLSAGPT LQYDKDRWLS TQWRLVSDEA VTNGLRDGIV FVLKCLDFSL VVNVKKIPFI ILSEEFIDPK SHKFVLRLQS ETSV
Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KAI1 C-terminal interacting tetraspanin; Loop-tail protein 2 homolog; LPP2; Strabismus 2; Van Gogh-like protein 1; vang-like 1 (van gogh, Drosophila); Vang-like protein 1
Gene Aliases: KITENIN; LPP2; STB2; STBM2; VANGL1
UniProt ID: (Human) Q8TAA9
Entrez Gene ID: (Human) 81839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support