Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
Immunogen sequence: TDALRFTLC SHGVQAGDLH LSVSEVQSCR HFCNKIWNAL RFILNALGEK FVPQPAEELS PSSPMDAWIL SRLALAAQEC ERGFLTRELS LVTHALHHFW LHNLCDVYLE AVKPVLWHSP RPLGPPQVLF SCADLGLRLL APLMPFLAEE LWQRLPPRPG CPPAPSISVA PYPSACSLEH WRQPELERRF SRVQEVVQVL RALRATYQLT KARPRVLLQS SEPGDQGLFE AFLEPLGTLG YCGAVGLLPP GAAAPSGWAQ APLSDTAQVY MELQGLVDPQ IQLPLLAARR YKLQKQLDSL TARTPSEGEA GTQRQQKLSS LQLELSKLDK AASHLQQLMD EPPAPGSPEL (168-516 aa encoded by BC008844)
This gene encodes a mitochondrial aminoacyl-tRNA synthetase, which catalyzes the attachment of valine to tRNA(Val) for mitochondrial translation. Mutations in this gene cause combined oxidative phosphorylation deficiency-20, and are also associated with early-onset mitochondrial encephalopathies. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: valine tRNA ligase 2, mitochondrial (putative); Valine--tRNA ligase, mitochondrial; ValRS; valyl trna synthetase; Valyl-tRNA synthetase; valyl-tRNA synthetase like; Valyl-tRNA synthetase-like
Gene Aliases: COXPD20; KIAA1885; VALRS; VARS2; VARS2L; VARSL
UniProt ID: (Human) Q5ST30
Entrez Gene ID: (Human) 57176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support