Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
VASH2 is a angiogenesis inhibitor. It inhibits network formation by endothelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testicular tissue protein Li 222; Tubulinyl-Tyr carboxypeptidase 2; Vasohibin-2; Vasohibin-like protein
Gene Aliases: VASH2; VASHL
UniProt ID: (Human) Q86V25
Entrez Gene ID: (Human) 79805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support