Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20230; membrane protein of cholinergic synaptic vesicles; Mitofusin-binding protein; Protein MIB; Synaptic vesicle membrane protein VAT-1 homolog; VATI; vesicle amine transport protein 1 homolog (T californica); vesicle amine transport protein 1 homolog (T. californica)
Gene Aliases: VAT-1; VAT1; VATI
UniProt ID: (Human) B4DPX4, (Rat) Q3MIE4, (Mouse) Q62465
Entrez Gene ID: (Human) 10493, (Rat) 287721, (Mouse) 26949
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support