Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RGLTELVEFY QQNSLKDCFK SLDTTLQFPF KEPEKRTISR PAVGSTKYFG TAKARYDFCA RDRSELSLKE GDIIKILNKK GQQGWWRGEI YGRVGWFPAN YVEEDYSEYC
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: oncogene vav; Proto-oncogene vav; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav proto-oncogene
Gene Aliases: VAV; VAV1
UniProt ID: (Human) P15498
Entrez Gene ID: (Human) 7409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support