Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: Jurkat, Mouse brain.
Immunogen sequence: TKESGEYAIS IKYNNEAKHI KILTRDGFFH IAENRKFKSL MELVEYYKHH SLKEGFRTLD TTLQFPYKEP EHSAGQRGNR AGNSLLSPKV LGIAIARYDF C
This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors (GEFs) for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ40431; Guanine nucleotide exchange factor VAV3; vav 3 guanine nucleotide exchange factor; vav 3 oncogene; VAV-3
Gene Aliases: A530094I06Rik; AA986410; Idd18.1; VAV3
UniProt ID: (Human) Q9UKW4, (Mouse) Q9R0C8
Entrez Gene ID: (Human) 10451, (Mouse) 57257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support