Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Acts as a deubiquitinating enzyme. Necessary for VCP-mediated reassembly of Golgi stacks after mitosis. May play a role in VCP-mediated formation of transitional endoplasmic reticulum (tER). Mediates dissociation of the ternary complex containing STX5A, NSFL1C and VCP. Hydrolyzes 'Lys-11'- and 'Lys-48'-linked polyubiquitin chains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating protein VCPIP1; valosin containing protein (p97)/p47 complex interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein p135; Valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCP/p47 complex-interacting 135-kDa protein
Gene Aliases: 4932442A08; 5730421J18Rik; 5730538E15Rik; DUBA3; KIAA1850; mKIAA1850; VCIP135; VCPIP1
UniProt ID: (Human) Q96JH7, (Mouse) Q8CDG3
Entrez Gene ID: (Human) 80124, (Mouse) 70675
Molecular Function: cysteine protease protease protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support