Sequence of this protein is as follows: MVELPLRHPA LFKAIGVKPP RGILLYGPPG TGKTLIARAV ANETGAFFFL INGPEIMSKL AGESESNLRK AFEEAEKNAP AIIFIDELDA IAPKREKTHG EVERRIVSQL LTLMDGLKQR AHVIVMAATN RPNSIDPALR RFGRFDREVD IGIPDATGRL EILQIHTKNM KLADDVDLEQ VANETHGHVG ADLAALCSEA ALQAIRKKMD LIDLEDETID AEVMNSLAVT MDDFRWALSQ SNPSALRETV VEVPQVTWED IGGLEDVKRE LQELVQYPVE HPDKFLKFGM TPSKGVLFYG PPGCGKTLLA KAIANECQAN FISIKGPELL TMWFGESEAN VREIFDKARQ AAPCVLFFDE LDSIAKARGG NIGDGGGAAD RVINQILTEM DGMSTKKNVF IIGATNRPDI IDPAILRPGR LDQLIYIPLP DEKSRVAILK ANLRKSPVAK DVDLEFLAKM TNGFSGADLT EICQRACKLA IRESIESEIR RERERQTNPS AMEVEEDDPV PEIRRDHFEE AMRFARRSVS DNDIRKYEMF AQTLQQSRGF GSFRFPSGNQ GGAGPSQGSG GGTGGSVYTE DNDDDLYG
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15S Mg(2+)-ATPase p97 subunit; AAA ATPase p97; epididymis luminal protein 220; epididymis secretory protein Li 70; homolog of yeast cdc48; Inv protein; or HEL-S-70; p97 atpase; TER ATPase; Transitional endoplasmic reticulum ATPase; Valosin-containing protein; VCP; yeast Cdc48p homolog
Gene Aliases: 3110001E05; ALS14; CDC48; CMT2Y; HEL-220; HEL-S-70; IBMPFD; IBMPFD1; p97; p97/VCP; TERA; VCP
UniProt ID: (Human) P55072, (Mouse) Q01853, (Rat) P46462
Entrez Gene ID: (Human) 7415, (Mouse) 269523, (Rat) 116643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support