Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSPKPRASGP PAKATEAGKR KSSSQPSPSD PKKKTTKVAK KGKAVRRGRR GKKGAATKMA AVTAPEAESG PAAPGPSDQP SQELPQHELP PEEPVSEGTQ HDPLSQESEL EEPLSQESEV EEPLSQESQV EEPLSQESEV EEPLSQESQV EEPLSQESEM EELPSM
May mediate a process in spermatogenesis or may play a role in sex ratio distortion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Variable charge protein on X with eight repeats; Variable charge X-linked protein 3; Variably charged protein X-A; variably charged X-A; VCX-8r; VCX-A
Gene Aliases: VCX-8r; VCX-A; VCX3; VCX3A; VCX8R; VCXA
UniProt ID: (Human) Q9NNX9
Entrez Gene ID: (Human) 51481
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support