Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Mouse: 98% identity (279/283 amino acids identical). Rat: 98% identity (279/283 amino acids identical) >60% identity with VDAC2 and VDAC3.
1 µg/mL of MA5-45666 was sufficient for detection of VDAC1 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 30kDa. Does not cross-react with VDAC2 or VDAC3 (based on KO validation results).
This antibody was formerly sold as clone S152B-23.
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hVDAC1; MGC111064; Mitochondrial Porin; mVDAC1; mVDAC5; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; Porin 31HL; Porin 31HM; PORIN-31-HL; rVDAC1; VD; VDAC-1; VDAC-5; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5
Gene Aliases: AL033343; PORIN; VDAC; VDAC-1; VDAC1; Vdac5
UniProt ID: (Human) P21796, (Mouse) Q60932, (Rat) Q9Z2L0
Entrez Gene ID: (Human) 7416, (Mouse) 22333, (Rat) 83529
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support