Product References
Morphogenetic events influencing corneal maturation, development, and transparency: Light and electron microscopic study.
Microscopy research and technique
Ibrahim N,Hifny A,Elhanbaly R,El-Desoky SMM,Gaber W
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Immunohistochemistry
Mon May 01 00:00:00 UTC 2023
Small extracellular vesicle-loaded bevacizumab reduces the frequency of intravitreal injection required for diabetic retinopathy.
Theranostics
Reddy SK,Ballal AR,Shailaja S,Seetharam RN,Raghu CH,Sankhe R,Pai K,Tender T,Mathew M,Aroor A,Shetty AK,Adiga S,Devi V,Muttigi MS,Upadhya D
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Wed May 10 00:00:00 UTC 2023
Human umbilical cord mesenchymal stem cells-derived exosomal circDLGAP4 promotes angiogenesis after cerebral ischemia-reperfusion injury by regulating miR-320/KLF5 axis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Feng J,He W,Xia J,Huang Q,Yang J,Gu WP,Zhang N,Liu YH
MA1-16629 was used in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot to find that exosomal circDLGAP4 negatively regulated miR-320 in HBMECs, which directly bound to KLF5. In addition, the downregulation of miR-320 could reverse the regulatory effect of exosomal shcircDLGAL5 in H/R-treated HBMECs by upregulating KLF5.
Wed Mar 01 00:00:00 UTC 2023
A Novel Preclinical In Vitro 3D Model of Oral Carcinogenesis for Biomarker Discovery and Drug Testing.
International journal of molecular sciences
Chitturi Suryaprakash RT,Shearston K,Farah CS,Fox SA,Iqbal MM,Kadolsky U,Zhong X,Saxena A,Kujan O
MA1-16629 was used in Immunohistochemistry to establish a 3D spheroid model of oral carcinogenesis for biomarker discovery and drug testing.
Fri Feb 17 00:00:00 UTC 2023
l-Citrulline ameliorates pathophysiology in a rat model of superimposed preeclampsia.
British journal of pharmacology
Man AWC,Zhou Y,Lam UDP,Reifenberg G,Werner A,Habermeier A,Closs EI,Daiber A,Münzel T,Xia N,Li H
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Wed Jun 01 00:00:00 UTC 2022
Characterization of the primo vascular system in rabbit vagina.
Microscopy research and technique
Mustafa FEA,Abdelhafez EA,Abd-Elhafeez HH
MA1-16629 was used in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot to characterize primo node as observed on the lymph vessel of rabbit vagina.
Tue Feb 01 00:00:00 UTC 2022
Influence of genistein and diadizine on regularity of estrous cycle in cyclic female Wistar rat: interaction with estradiol receptors and vascular endothelial growth factor.
Open veterinary journal
Elsayed DH,Helmy SA,Dessouki AA,El-Nahla AM,Abdelrazek HMA,El-Hak HNG
MA1-16629 was used in Immunohistochemistry-paraffin to investigate the interactions of isoflavones with estrogen receptors alpha (ERα), beta (ERβ), and vascular endothelial growth factor (VEGF) expressions in ovarian and uterine tissues during different stages of the estrous cycle of regular cyclic female Wistar rats.
Tue Jan 03 00:00:00 UTC 2023
Characterization of giant neuroendocrine cells in the pregnant New Zealand white rabbit vagina demonstrated by histological, histochemical, and immunohistochemical methods.
Microscopy research and technique
Mustafa FEA
MA1-16629 was used in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot to detect the presence of giant neuroendocrine cells in the vagina of the pregnant rabbit.
Sat Oct 01 00:00:00 UTC 2022
Impact of methionine restriction on muscle aerobic metabolism and hypertrophy in young and old mice on an obesogenic diet.
Growth factors (Chur, Switzerland)
Swaminathan A,Cesanelli L,Venckunas T,Degens H
MA1-16629 was used in Western Blot to study the effects of MR (0.17% kCal methionine, 10% kCal fat) and MR + high-fat diet (HFD) (0.17% methionine, 45% kCal fat) and overload-induced hypertrophy on inflammation, angiogenesis and mitochondrial activity in the hind-limb muscle in 10- and 26-month-old male C57BL/6J mice.
Mon Aug 01 00:00:00 UTC 2022
The RNA Methyltransferase METTL3 Promotes Endothelial Progenitor Cell Angiogenesis in Mandibular Distraction Osteogenesis via the PI3K/AKT Pathway.
Frontiers in cell and developmental biology
Jiang W,Zhu P,Huang F,Zhao Z,Zhang T,An X,Liao F,Guo L,Liu Y,Zhou N,Huang X
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Fri Nov 19 00:00:00 UTC 2021
Targeting EYA3 in Ewing Sarcoma Retards Tumor Growth and Angiogenesis.
Molecular cancer therapeutics
Wang Y,Pandey RN,Roychoudhury K,Milewski D,Kalin TV,Szabo S,Pressey JG,Hegde RS
MA1-16629 was used in Immunohistochemistry to investigate molecular and cellular mechanisms through which EYA3 might promote Ewing sarcoma tumor growth and to determine whether the EYA3 tyrosine phosphatase activity represents a viable therapeutic target.
Sat May 01 00:00:00 UTC 2021
Targeting EYA3 in Ewing Sarcoma Retards Tumor Growth and Angiogenesis.
Molecular cancer therapeutics
Wang Y,Pandey RN,Roychoudhury K,Milewski D,Kalin TV,Szabo S,Pressey JG,Hegde RS
MA1-16629 was used in Immunohistochemistry to investigate molecular and cellular mechanisms through which EYA3 might promote Ewing sarcoma tumor growth and to determine whether the EYA3 tyrosine phosphatase activity represents a viable therapeutic target.
Sat May 01 00:00:00 UTC 2021
An Injectable Fibrin Scaffold Rich in Growth Factors for Skin Repair.
BioMed research international
Shao Z,Lyu C,Teng L,Xie X,Sun J,Zou D,Lu J
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Immunohistochemistry
Tue May 25 00:00:00 UTC 2021
Physiological Effects of Intermittent Passive Exposure to Hypobaric Hypoxia and Cold in Rats.
Frontiers in physiology
Santocildes G,Viscor G,Pagès T,Ramos-Romero S,Torres JL,Torrella JR
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Sat Jun 19 00:00:00 UTC 2021
Physiological Effects of Intermittent Passive Exposure to Hypobaric Hypoxia and Cold in Rats.
Frontiers in physiology
Santocildes G,Viscor G,Pagès T,Ramos-Romero S,Torres JL,Torrella JR
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Sat Jun 19 00:00:00 UTC 2021
MicroRNA-205 mediates endothelial progenitor functions in distraction osteogenesis by targeting the transcription regulator NOTCH2.
Stem cell research & therapy
Jiang W,Zhu P,Zhang T,Liao F,Yu Y,Liu Y,Shen H,Zhao Z,Huang X,Zhou N
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Wed Feb 03 00:00:00 UTC 2021
Circ_CLASP2 Regulates High Glucose-Induced Dysfunction of Human Endothelial Cells Through Targeting miR-140-5p/FBXW7 Axis.
Frontiers in pharmacology
Zhang Q,Long J,Li N,Ma X,Zheng L
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Tue Mar 30 00:00:00 UTC 2021
Circ_CLASP2 Regulates High Glucose-Induced Dysfunction of Human Endothelial Cells Through Targeting miR-140-5p/FBXW7 Axis.
Frontiers in pharmacology
Zhang Q,Long J,Li N,Ma X,Zheng L
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Western Blot
Tue Mar 30 00:00:00 UTC 2021
Differential expression of angiogenesis markers HSP70, HSP90, VEGF and pERK1/2 in both components of dedifferentiated chondrosarcomas.
Journal of bone oncology
Schoedel K,Miller V,Osei-Hwedieh D,Watters R,Duensing A,John I,Chandran U,Chang A,Soman V,Weiss K
MA1-16629 was used in Immunohistochemistry to study the expression of angiogenesis markers as future treatment targets for dedifferentiated chondrosarcomas.
Sun Aug 01 00:00:00 UTC 2021
Differential expression of angiogenesis markers HSP70, HSP90, VEGF and pERK1/2 in both components of dedifferentiated chondrosarcomas.
Journal of bone oncology
Schoedel K,Miller V,Osei-Hwedieh D,Watters R,Duensing A,John I,Chandran U,Chang A,Soman V,Weiss K
MA1-16629 was used in Immunohistochemistry to study the expression of angiogenesis markers as future treatment targets for dedifferentiated chondrosarcomas.
Sun Aug 01 00:00:00 UTC 2021
Effect of soy isoflavones on implantation losses in Wistar rat: implication of progesterone receptors, vascular endothelial growth factor and estradiol receptors alpha.
Iranian journal of veterinary research
Elsayed DH,Abdelrazek HMA,Eltamany DA,Ebaid HM,El-Nahla AM
MA1-16629 was used in Immunohistochemistry to study the implantation losses caused by soy isoflavones.
Thu Apr 14 00:00:00 UTC 2022
Effect of vitamin B12 supplementation on retinal lesions in diabetic rats.
Molecular vision
Reddy SS,Prabhakar YK,Kumar CU,Reddy PY,Reddy GB
MA1-16629 was used in Western Blotting to investigate the impact of dietary supplementation of vitamin B12 on retinal changes in diabetic rats.
Mon May 10 00:00:00 UTC 2021
Effect of vitamin B12 supplementation on retinal lesions in diabetic rats.
Molecular vision
Reddy SS,Prabhakar YK,Kumar CU,Reddy PY,Reddy GB
MA1-16629 was used in Western Blotting to investigate the impact of dietary supplementation of vitamin B12 on retinal changes in diabetic rats.
Mon May 10 00:00:00 UTC 2021
An updated investigation on the dromedary camel cerebellum (Camelus dromedarius) with special insight into the distribution of calcium-binding proteins.
Scientific reports
Attaai AH,Noreldin AE,Abdel-Maksoud FM,Hussein MT
MA1-16629 was used in Immunohistochemistry (Paraffin) to display a complete map for the neurochemical organization of CaBP expression and distribution by different cells in the camel cerebellum.
Thu Dec 03 00:00:00 UTC 2020
An updated investigation on the dromedary camel cerebellum (Camelus dromedarius) with special insight into the distribution of calcium-binding proteins.
Scientific reports
Attaai AH,Noreldin AE,Abdel-Maksoud FM,Hussein MT
MA1-16629 was used in Immunohistochemistry (Paraffin) to display a complete map for the neurochemical organization of CaBP expression and distribution by different cells in the camel cerebellum.
Thu Dec 03 00:00:00 UTC 2020
Contractile Activity Is Necessary to Trigger Intermittent Hypobaric Hypoxia-Induced Fiber Size and Vascular Adaptations in Skeletal Muscle.
Frontiers in physiology
Rizo-Roca D,Bonet JB,Ínal B,Ríos-Kristjánsson JG,Pagès T,Viscor G,Torrella JR
MA1-16629 was used in Western Blotting to study the effects of intermittent hypobaric hypoxia on different myofiber morphofunctional parameters and investigate whether contractile activity is required to elicit hypoxia-induced adaptations in trained rats.
Wed Sep 30 00:00:00 UTC 2020
Contractile Activity Is Necessary to Trigger Intermittent Hypobaric Hypoxia-Induced Fiber Size and Vascular Adaptations in Skeletal Muscle.
Frontiers in physiology
Rizo-Roca D,Bonet JB,Ínal B,Ríos-Kristjánsson JG,Pagès T,Viscor G,Torrella JR
MA1-16629 was used in Western Blotting to study the effects of intermittent hypobaric hypoxia on different myofiber morphofunctional parameters and investigate whether contractile activity is required to elicit hypoxia-induced adaptations in trained rats.
Wed Sep 30 00:00:00 UTC 2020
Topical Loperamide-Encapsulated Liposomal Gel Increases the Severity of Inflammation and Accelerates Disease Progression in the Adjuvant-Induced Model of Experimental Rheumatoid Arthritis.
Frontiers in pharmacology
Hua S,Dias TH,Pepperall DG,Yang Y
Published figure using VEGF monoclonal antibody (Product # MA1-16629) in Immunohistochemistry
Wed Sep 30 00:00:00 UTC 2020
The Eyes Absent Proteins in Developmental and Pathological Angiogenesis.
The American journal of pathology
Wang Y,Tadjuidje E,Pandey RN,Stefater JA,Smith LE,Lang RA,Hegde RS
MA1-16629 was used in immunocytochemistry to study proteins absent in the eyes during pathological and developmental angiogenesis
Tue Mar 01 00:00:00 UTC 2016
Comparative study of membranes induced by PMMA or silicone in rats, and influence of external radiotherapy.
Acta biomaterialia
de Monès E,Schlaubitz S,Oliveira H,d'Elbée JM,Bareille R,Bourget C,Couraud L,Fricain JC
MA1-16629 was used in western blot to investigate the benefits of replacing a PMMA spacer with a semi-flexible medical grade silicone spacer for long bone defect reconstruction
Mon Jun 01 00:00:00 UTC 2015
Gastric cancer in young vs old Romanian patients: immunoprofile with emphasis on maspin and mena protein reactivity.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Gurzu S,Kadar Z,Sugimura H,Bara T,Bara T,Halmaciu I,Jung I
MA1-16629 was used in immunohistochemistry - paraffin section to use Romanian patients with gastric carcinomas and compare the demographic, clinical, and immunohistochemical aspects of each cancer
Sun Mar 01 00:00:00 UTC 2015
Oxygen-induced retinopathy in mice with retinal photoreceptor cell degeneration.
Life sciences
Zhang Q,Zhang ZM
MA1-16629 was used in western blot to study retinal neovascularization during oxygen-induced retinopathy in mice with photoreceptor cell degeneration
Fri Apr 25 00:00:00 UTC 2014
Thymic epithelial tumors express vascular endothelial growth factors and their receptors as potential targets of antiangiogenic therapy: a tissue micro array-based multicenter study.
Lung cancer (Amsterdam, Netherlands)
Lattanzio R,La Sorda R,Facciolo F,Sioletic S,Lauriola L,Martucci R,Gallo E,Palmieri G,Evoli A,Alessandrini G,Ruco L,Rendina EA,Truini M,Chiarle R,Barreca A,Pich A,Ascani S,Remotti D,Tunesi G,Granone P,Ratto GB,Puma F,Pescarmona E,Piantelli M,Marino M,Carlini S,Cerasoli V,Corzani F,Melis E,Filippetti M,Canalini P,Palestro G,Lalle M,Ruffini E,Ceribelli A,Rinaldi M
MA1-16629 was used in immunohistochemistry to study the immunohistochemical expression of VEGF and VEGFR isoforms in patients with thymic epithelial tumors
Fri Aug 01 00:00:00 UTC 2014
The effect of lipoic acid on wound healing in a full thickness uterine injury model in rats.
Journal of molecular histology
Micili SC,Goker A,Sayin O,Akokay P,Ergur BU
MA1-16629 was used in immunohistochemistry to use a rat full thickness uterine injury model to study the beneficial effects of lipoic acid on wound healing
Sat Jun 01 00:00:00 UTC 2013
Paraneoplastic symptoms: cachexia, polycythemia, and hypercalcemia are, respectively, related to vascular endothelial growth factor (VEGF) expression in renal clear cell carcinoma.
Urologic oncology
Ding GX,Feng CC,Song NH,Fang ZJ,Xia GW,Jiang HW,Hua LX,Ding Q
MA1-16629 was used in immunohistochemistry to study the relationship between expression of VEGF and the development of paraneoplastic symptoms in patients with renal clear cell carcinoma
Fri Nov 01 00:00:00 UTC 2013
The efficacy of bevacizumab, sorafenib, and retinoic acid on rat endometriosis model.
Reproductive sciences (Thousand Oaks, Calif.)
Ozer H,Boztosun A,Açmaz G,Atilgan R,Akkar OB,Kosar MI
MA1-16629 was used in immunohistochemistry to compare the therapeutic effects of bevacizumab, sorafenib and nicotinic acid in a rat model of endometriosis
Tue Jan 01 00:00:00 UTC 2013
Resveratrol treatment delays growth plate fusion and improves bone growth in female rabbits.
PloS one
Karimian E,Tamm C,Chagin AS,Samuelsson K,Kjartansdóttir KR,Ohlsson C,Sävendahl L
MA1-16629 was used in immunohistochemistry to study the potential mechanisms involved in the delayed growth plate fusion and increased longitudonal bome growth observed in female rabbits treated with resveratrol
Thu Apr 24 00:00:00 UTC 2014
Mesenchymal-to-endothelial transition in Kaposi sarcoma: a histogenetic hypothesis based on a case series and literature review.
PloS one
Gurzu S,Ciortea D,Munteanu T,Kezdi-Zaharia I,Jung I
MA1-16629 was used in immunohistochemistry to develop a hypothesis of Kaposi sarcoma histogenesis based on a case and literature review
Tue Apr 01 00:00:00 UTC 2014
The relevance of the intrinsic subtype to the clinicopathological features and biomarkers in Japanese breast cancer patients.
World journal of surgical oncology
Tamaki M,Kamio T,Kameoka S,Kojimahara N,Nishikawa T
MA1-16629 was used in immunohistochemistry to study the effect of breast cancer intrinsic subtype on cliniopathological and biomarker profiles in Japanese patients
Mon Nov 18 00:00:00 UTC 2013
The impact of KRAS mutations on VEGF-A production and tumour vascular network.
BMC cancer
Figueras A,Arbos MA,Quiles MT,Viñals F,Germà JR,Capellà G
MA1-16629 was used in western blot to study the differential effects on the production of VEGF-A and tumor vasculature development of codon 12 and codon 13 KRAS mutations
Mon Mar 18 00:00:00 UTC 2013
Temporal and pharmacological characterization of angiostatin release and generation by human platelets: implications for endothelial cell migration.
PloS one
Radziwon-Balicka A,Moncada de la Rosa C,Zielnik B,Doroszko A,Jurasz P
MA1-16629 was used in immunocytochemistry to study the production and release of angiostatin by human platelets
Tue Sep 24 00:00:00 UTC 2013
Diagnostic and therapeutic strategy and the most efficient prognostic factors of breast malignant fibrous histiocytoma.
Scientific reports
Qiu SQ,Wei XL,Huang WH,Wu MY,Qin YS,Li YK,Zhang GJ
MA1-16629 was used in immunohistochemistry - paraffin section to report the clinicopathological features of 9 breast malignant fibrous histiocytoma patients
Fri Feb 14 00:00:00 UTC 2014
Paraneoplastic hormones: parathyroid hormone-related protein (PTHrP) and erythropoietin (EPO) are related to vascular endothelial growth factor (VEGF) expression in clear cell renal cell carcinoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Feng CC,Ding GX,Song NH,Li X,Wu Z,Jiang HW,Ding Q
MA1-16629 was used in immunohistochemistry to study the expression of PTHrP, EPO and VEGF in clear cell renal cell carcinoma and their relationships with tumor grade
Sun Dec 01 00:00:00 UTC 2013
Biological and demographic profile of meningiomas in a cohort of Egyptian patients: impact on tumor recurrence.
TheScientificWorldJournal
Abdelzaher E,El Deeb NM,Gowil AG,Yehya A
MA1-16629 was used in immunohistochemistry to study the relationship between meningioma biological and demographic profiles and disease recurrence in Egyptian patients
Mon Jun 16 00:00:00 UTC 2014
The role of thrombin receptors PAR1 and PAR4 for PAI-1 storage, synthesis and secretion by human platelets.
Thrombosis research
Nylander M,Osman A,Ramström S,Aklint E,Larsson A,Lindahl TL
MA1-16629 was used in immunocytochemistry to study the production and secretion of PAI-1 by human platelets and the roles played by the the PAR1 and PAR4 thrombin receptors
Sun Apr 01 00:00:00 UTC 2012
Prediction of the shrinking rate of uterine leiomyoma nodules using needle biopsy specimens.
Fertility and sterility
Kasai M,Ichimura T,Kawamura N,Sumikura T,Matsuda M,Asano C,Sumi T,Ishiko O
MA1-16629 was used in immunohistochemistry to study the relationship between the shrinking of uterine leiomyoma nodules and the expression of ER, PR and VEGF following menopause or pharmacological treatment
Wed Aug 01 00:00:00 UTC 2012
Boswellic acid inhibits growth and metastasis of human colorectal cancer in orthotopic mouse model by downregulating inflammatory, proliferative, invasive and angiogenic biomarkers.
International journal of cancer
Yadav VR,Prasad S,Sung B,Gelovani JG,Guha S,Krishnan S,Aggarwal BB
MA1-16629 was used in immunohistochemistry to study the mechanisms underlying the inhibitory effects of boswellic acid on human colorectal cancer growth and metasatsis in an orthotopic murine model
Tue May 01 00:00:00 UTC 2012
Apigenin attenuates insulin-like growth factor-I signaling in an autochthonous mouse prostate cancer model.
Pharmaceutical research
Shukla S,MacLennan GT,Fu P,Gupta S
MA1-16629 was used in western blot to study the effects of the plant flavone apigenin on IGF-1 signaling, angiogenesis, and metastasis in a murine prostate cancer model
Fri Jun 01 00:00:00 UTC 2012
Lipoxin A4 inhibits the development of endometriosis in mice: the role of anti-inflammation and anti-angiogenesis.
American journal of reproductive immunology (New York, N.Y. : 1989)
Xu Z,Zhao F,Lin F,Chen J,Huang Y
MA1-16629 was used in immunohistochemistry to study the anti-inflammatory and anti-angiogenic mechanisms by which lipoxin A4 inhibits endometriosis in a murine model
Fri Jun 01 00:00:00 UTC 2012
Bronchial carcinoid tumors metastatic to the sella turcica and review of the literature.
Pituitary
Moshkin O,Rotondo F,Scheithauer BW,Soares M,Coire C,Smyth HS,Goth M,Horvath E,Kovacs K
MA1-16629 was used in immunohistochemistry to report on a case of bronchial carcinoid tumors metastatic to the sella turcica
Fri Jun 01 00:00:00 UTC 2012
Immunohistochemical study of the angiogenetic network of VEGF, HIF1α, VEGFR-2 and endothelial nitric oxide synthase (eNOS) in human breast cancer.
Pathology oncology research : POR
Kafousi M,Vrekoussis T,Tsentelierou E,Pavlakis K,Navrozoglou I,Dousias V,Sanidas E,Tsiftsis D,Georgoulias V,Stathopoulos EN
MA1-16629 was used in immunohistochemistry to study the expression of VEGF, VEGFR-2, HIF1-alpha and eNOS in invasive ductal carcinoma
Sun Jan 01 00:00:00 UTC 2012
Pulmonary artery sarcoma with angiosarcoma phenotype mimicking pleomorphic malignant fibrous histiocytoma: a case report.
Diagnostic pathology
Bohn OL,de León EA,Lezama O,Rios-Luna NP,Sánchez-Sosa S,Llombart-Bosch A
MA1-16629 was used in immunohistochemistry to report on a case of pulmonary artery sarcoma
Wed Nov 07 00:00:00 UTC 2012
VEGFR1 expression is related to lymph node metastasis and serum VEGF may be a marker of progression in the follow-up of patients with differentiated thyroid carcinoma.
European journal of endocrinology
Karaca Z,Tanriverdi F,Unluhizarci K,Ozturk F,Gokahmetoglu S,Elbuken G,Cakir I,Bayram F,Kelestimur F
MA1-16629 was used in immunohistochemistry to investigate the roles of VEGFR1 and VEGF during lymph node metastasis and differentiated thyroid carcinoma
Tue Feb 01 00:00:00 UTC 2011
Effect of phototherapy on growth factor levels in neonatal rat skin.
Journal of pediatric surgery
Soyer T,Ayva S,Aliefendioğlu D,Aktuna Z,Aslan MK,Senyücel MF,Cakmak M
MA1-16629 was used in immunohistochemistry to study the effects of phototherapy on the levels of growth factors involved in wound healing in neonatal rat skin
Tue Nov 01 00:00:00 UTC 2011
Immunolocalization of VEGF, VEGF receptors, EGF-R and Ki-67 in leiomyoma, cellular leiomyoma and leiomyosarcoma.
Acta histochemica
Sanci M,Dikis C,Inan S,Turkoz E,Dicle N,Ispahi C
MA1-16629 was used in immunohistochemistry to study the role of VEGF, VEGF receptors, EGF receptor and Ki-67 in leiomyosarcoma
Sun May 01 00:00:00 UTC 2011
The effect of antivascular endothelial growth factor on the development of adhesion formation in laparotomized rats: experimental study.
Gastroenterology research and practice
Basbug M,Bulbuller N,Camci C,Ayten R,Aygen E,Ozercan IH,Arikanoglu Z,Akbulut S
MA1-16629 was used in immunohistochemistry to study the effect of an anti-VEGF monoclonal antibody on the formation of adhesions in the rat cecal abrasion model
Thu Nov 10 00:00:00 UTC 2011
Angiogenesis in the progression of breast ductal proliferations.
International journal of surgical pathology
Carpenter PM,Chen WP,Mendez A,McLaren CE,Su MY
MA1-16629 was used in immunohistochemistry to study the role of angiogenesis in the progression of breast ductal carcinomas
Wed Jun 01 00:00:00 UTC 2011
Serum and pulmonary vascular endothelial growth factor/receptors and haemodynamic measurements in cyanotic congenital heart disease with decreased pulmonary blood flow.
Cardiology in the young
Tolunay I,Tunaoglu S,Akyürek N,Halid V,Olgunturk R,Kula S
MA1-16629 was used in immunohistochemistry to study levels of VEGF and VEGFR in the serum and lungs of patients with cyanotic congenital heart disease
Thu Dec 01 00:00:00 UTC 2011
Vascular endothelial growth factor (VEGF) enhances gastric carcinoma invasiveness via integrin alpha(v)beta6.
Cancer letters
Zhao R,Liu XQ,Wu XP,Liu YF,Zhang ZY,Yang GY,Guo S,Niu J,Wang JY,Xu KS
MA1-16629 was used in immunohistochemistry to investigate the effect of vascular endothelial growth factor on gastric carcinoma invasiveness and its mechanism
Thu Jan 28 00:00:00 UTC 2010
Influence of 670 nm low-level laser therapy on mast cells and vascular response of cutaneous injuries.
Journal of photochemistry and photobiology. B, Biology
Pereira MC,de Pinho CB,Medrado AR,Andrade Zde A,Reis SR
MA1-16629 was used in immunohistochemistry to study the effect laser therapy on the inflammatory and vascular responses of cutaneous wounds
Mon Mar 08 00:00:00 UTC 2010
Prevention of ovarian hyperstimulation syndrome in a rat model: efficacy comparison between cabergoline and meloxicam.
Acta obstetricia et gynecologica Scandinavica
Saylan A,Arioz DT,Koken T,Dilek H,Saylan F,Yilmazer M
MA1-16629 was used in immunohistochemistry to compare the effectiveness between cabergoline and meloxicam in the prevention of ovarian hyperstimulation syndrome
Sat May 01 00:00:00 UTC 2010
Anti-human vascular endothelial growth factor (VEGF) antibody selection for immunohistochemical staining of proliferating blood vessels.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
van der Loos CM,Meijer-Jorna LB,Broekmans ME,Ploegmakers HP,Teeling P,de Boer OJ,van der Wal AC
MA1-16629 was used in immunohistochemistry to evaluate suitable vascular endothelial growth factor antibodies for the detection of proliferating blood vessels
Mon Feb 01 00:00:00 UTC 2010
Vascular endothelial growth factor: a possible mediator of endothelial activation in acute respiratory distress syndrome.
Minerva anestesiologica
Azamfirei L,Gurzu S,Solomon R,Copotoiu R,Copotoiu S,Jung I,Tilinca M,Branzaniuc K,Corneci D,Szederjesi J,Kovacs J
MA1-16629 was used in immunohistochemistry to study the potential role of VEGF in mediating endothelial activation in acute respiratory distress syndrome
Sun Aug 01 00:00:00 UTC 2010
Molecular biomarkers for advanced renal cell carcinoma: implications for prognosis and therapy.
Urologic oncology
Kim HS,Kim WS,Park SH,Jung CW,Choi HY,Lee HM,Jeon SS,Ha H,Hwang IG,Lee S,Lim HY
MA1-16629 was used in immunohistochemistry to identify reliable predictive biomarkers for patients with metastatic renal cell carcinomas
Mon Jun 14 00:00:00 UTC 2010
Maspin, VEGF and p53 expression in small biopsies of primary advanced lung cancer and relationship with clinicopathologic parameters.
Pathology oncology research : POR
Bircan A,Bircan S,Kapucuoglu N,Songur N,Ozturk O,Akkaya A
MA1-16629 was used in immunohistochemistry to compare expression of Maspin, VEGF, and p53 between carcinomas
Wed Dec 01 00:00:00 UTC 2010
Evaluation of relationship between HIF-1alpha immunoreactivity and stage, grade, angiogenic profile and proliferative index in bladder urothelial carcinomas.
International urology and nephrology
Deniz H,Karakök M,Yagci F,Güldür ME
MA1-16629 was used in immunohistochemistry to study the relationship between HIF-1alpha expression and various prognostic parameters in bladder urothelial carcinomas
Mon Mar 01 00:00:00 UTC 2010
Podocyte-specific overexpression of GLUT1 surprisingly reduces mesangial matrix expansion in diabetic nephropathy in mice.
American journal of physiology. Renal physiology
Zhang H,Schin M,Saha J,Burke K,Holzman LB,Filipiak W,Saunders T,Xiang M,Heilig CW,Brosius FC
MA1-16629 was used in western blot to investigate the effect of the overexpression of GLUT1 in podocytes on the mesangial matrix expansion in rodent diabetic nephropathy
Thu Jul 01 00:00:00 UTC 2010
Clinical and immunohistochemical features associated with a response to bortezomib in patients with multiple myeloma.
Clinical cancer research : an official journal of the American Association for Cancer Research
Dawson MA,Opat SS,Taouk Y,Donovan M,Zammit M,Monaghan K,Horvath N,Roberts AW,Prince HM,Hertzberg M,McLean CA,Spencer A
MA1-16629 was used in immunohistochemistry - paraffin section to investigate the parameters for the identification of multiple myeloma patients suitable for bortezomib therapy
Thu Jan 15 00:00:00 UTC 2009
Bevacizumab and breast cancer: current therapeutic progress and future perspectives.
Expert review of anticancer therapy
Yang SX
MA1-16629 was used in immunohistochemistry to evaluate the efficacy of bevacizumab treatment for breast cancer
Tue Dec 01 00:00:00 UTC 2009
The differences between the endothelial area marked with CD31 and CD105 in colorectal carcinomas by computer-assisted morphometrical analysis.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Jung I,Gurzu S,Raica M,Cîmpean AM,Szentirmay Z
MA1-16629 was used in immunohistochemistry to compare the CD31 and CD105 endothelial areas in colorectal carcinomas by computer-assisted morphometrical analysis
Fri Sep 18 00:00:00 UTC 2009
Association of vascular endothelial growth factor and vascular endothelial growth factor receptor-2 genetic polymorphisms with outcome in a trial of paclitaxel compared with paclitaxel plus bevacizumab in advanced breast cancer: ECOG 2100.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Schneider BP,Wang M,Radovich M,Sledge GW,Badve S,Thor A,Flockhart DA,Hancock B,Davidson N,Gralow J,Dickler M,Perez EA,Cobleigh M,Shenkier T,Edgerton S,Miller KD
MA1-16629 was used in immunohistochemistry to investigate the prognostic significance of vascular endothelial growth factor and vascular endothelial growth factor receptor-2 genetic polymorphisms in breast cancer treatment
Wed Oct 01 00:00:00 UTC 2008
Neoangiogenesis and expression of hypoxia-inducible factor 1alpha, vascular endothelial growth factor, and glucose transporter-1 in endometrioid type endometrium adenocarcinomas.
Gynecologic oncology
Ozbudak IH,Karaveli S,Simsek T,Erdogan G,Pestereli E
MA1-16629 was used in immunohistochemistry to study the expression of HIF-1alpha, VEGF and GLUT-1 in endometrioid type endometrium adenocarcinomas
Sat Mar 01 00:00:00 UTC 2008
Vitreous fluid of db/db mice exhibits alterations in angiogenic and metabolic factors consistent with early diabetic retinopathy.
Ophthalmic research
Cohen MP,Hud E,Shea E,Shearman CW
MA1-16629 was used in western blot to study whether angiogenic and metabolic factors in vitreous fluid of db/db mice reflect early diabetic retinopathy
Tue Jan 15 00:00:00 UTC 2008
Vascular endothelial growth factor expression and vascularity in renal allograft biopsies.
Transplantation proceedings
Sarioğlu S,Celik A,Erşen A,Uçer I,Sağlam F,Camsari T,Gülay H
MA1-16629 was used in immunohistochemistry to investigate the relationship between vascular endothelial growth factor expression and the pathology in renal allograft biopsies
Thu May 01 00:00:00 UTC 2008
The expression of cytoskeleton regulatory protein Mena in colorectal lesions.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Gurzu S,Jung I,Prantner I,Ember I,Pávai Z,Mezei T
MA1-16629 was used in immunohistochemistry to investigate the expression of mammalian actin regulatory protein Ena in colorectal lesions
Thu Dec 11 00:00:00 UTC 2008
Effects of stobadine and vitamin E in diabetes-induced retinal abnormalities: involvement of oxidative stress.
Archives of medical research
Yülek F,Or M,Ozoğul C,Isik AC,Ari N,Stefek M,Bauer V,Karasu C
MA1-16629 was used in immunohistochemistry to investigate the therapeautical benefits of stobadine and vitamin E in diabetes-induced retinal abnormalities
Sun Jul 01 00:00:00 UTC 2007
Prognostic significance of HIF-2alpha/EPAS1 expression in hepatocellular carcinoma.
World journal of gastroenterology
Bangoura G,Liu ZS,Qian Q,Jiang CQ,Yang GF,Jing S
MA1-16629 was used in immunohistochemistry to investigate the role of the HIF-2alpha/EPAS1 in hepatocellular carcinoma
Thu Jun 21 00:00:00 UTC 2007
Angiogenesis in nodal B cell lymphomas: a high throughput study.
Journal of clinical pathology
Tzankov A,Heiss S,Ebner S,Sterlacci W,Schaefer G,Augustin F,Fiegl M,Dirnhofer S
MA1-16629 was used in immunohistochemistry to study angiogenesis in B cell lymphomas
Tue May 01 00:00:00 UTC 2007
Vascular endothelial growth factor, hypoxia-inducible factor 1 alpha and CD34 expressions in early-stage gastric tumors: relationship with pathological factors and prognostic impact on survival.
Oncology
Cabuk D,Basaran G,Celikel C,Dane F,Yumuk PF,Iyikesici MS,Ekenel M,Turhal NS
MA1-16629 was used in immunohistochemistry to evaluate the prognostic markers for early-stage gastric tumors
Wed Jan 02 00:00:00 UTC 2008
Immunohistochemical analysis with multiple antibodies in search of prognostic markers for clear cell renal cell carcinoma.
Urology
Phuoc NB,Ehara H,Gotoh T,Nakano M,Yokoi S,Deguchi T,Hirose Y
MA1-16629 was used in immunohistochemistry to identify prognostic markers for clear cell renal cell carcinoma
Tue May 01 00:00:00 UTC 2007
Evaluation of the prognostic role of a panel of biomarkers in stage IB-IIIA non-small cell lung cancer patients.
Anticancer research
Timotheadou E,Skarlos DV,Samantas E,Papadopoulos S,Murray S,Skrickova J,Christodoulou C,Papakostantinou C,Pectasides D,Papakostas P,Kaplanova J,Vrettou E,Karina M,Kosmidis P,Fountzilas G
MA1-16629 was used in immunohistochemistry to investigate the prognostic biomarkers in stage IB-IIIA non-small cell lung cancer
Thu Feb 28 00:00:00 UTC 2008
Effects of finasteride on the vascular surface density, number of microvessels and vascular endothelial growth factor expression of the rat prostate.
International urology and nephrology
Canda AE,Mungan MU,Yilmaz O,Yorukoglu K,Tuzel E,Kirkali Z
MA1-16629 was used in immunohistochemistry to study the effects of a 5-alpha reductase inhibitor on vascularity and VEGF expression in the rat prostate
Mon Mar 05 00:00:00 UTC 2007
Microvessel density, VEGF expression, and tumor-associated macrophages in breast tumors: correlations with prognostic parameters.
Journal of experimental & clinical cancer research : CR
Bolat F,Kayaselcuk F,Nursal TZ,Yagmurdur MC,Bal N,Demirhan B
MA1-16629 was used in immunohistochemistry to study the relationship between microvessel density, VEGF expression, and tumor-associated macrophages in breast tumors
Fri Sep 01 00:00:00 UTC 2006
PGE1 analog alprostadil induces VEGF and eNOS expression in endothelial cells.
American journal of physiology. Heart and circulatory physiology
Haider DG,Bucek RA,Giurgea AG,Maurer G,Glogar H,Minar E,Wolzt M,Mehrabi MR,Baghestanian M
MA1-16629 was used in western blot to study the role of the PGE1 analog alprostadil in inducing VEGF and eNOS expression in endothelial cells
Tue Nov 01 00:00:00 UTC 2005