Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
The target sequence has 100% sequence homology with mouse and rat.
Receptors for VEGF, VEGFB and PGF have a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. VEGF and its high-affinity binding receptors, the tyrosine kinases FLK1 and FLT1, are thought to be important for the development of embryonic vasculature. It has been shown that an alternately spliced form of FLT1 produces a soluble protein, termed sFLT1, which binds vascular endothelial growth factor with high affinity, playing an inhibitory role in angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD309; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; FLT; FLT-1; Fms-like tyrosine kinase 1; fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); KDR; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Protein-tyrosine kinase receptor flk-1; soluble VEGFR 1; soluble VEGFR2; sVEGFR 1; tyrosine kinase growth factor receptor; Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; Vascular endothelial growth factor receptor 2; Vascular permeability factor receptor; VEGF R1; VEGFR 1; VEGFR-1; VEGFR-2
Gene Aliases: CD309; FLK1; FLT; FLT-1; FLT1; FRT; KDR; VEGFR; VEGFR-1; VEGFR1; VEGFR2
UniProt ID: (Human) P17948, (Human) P35968
Entrez Gene ID: (Human) 2321, (Human) 3791
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support