Product References
Transplantation of Human Amniotic Mesenchymal Stem Cells Up-Regulates Angiogenic Factor Expression to Attenuate Diabetic Kidney Disease in Rats.
Diabetes, metabolic syndrome and obesity : targets and therapy
Ni Y,Chen Y,Jiang X,Pu T,Zhang L,Li S,Hu L,Bai B,Hu T,Yu L,Yang Y
Wed Feb 15 00:00:00 UTC 2023
Protocol for preparation of a photo-triggering double cross-linked adhesive, antibacterial, and biocompatible hydrogel for wound healing.
STAR protocols
Zhai X,Hu M,Hu H,Yao X,Wei W
Sat May 13 00:00:00 UTC 2023
LncRNA SPRY4-IT1 facilitates cell proliferation and angiogenesis of glioma via the miR-101-3p/EZH2/VEGFA signaling axis.
Cancer medicine
Wang J,Chen Y,Wang Q,Xu H,Jiang Q,Wang M,Li S,Chen Y,Wu C,Yu P,Xiao Z,Chen W,Lan Q
Wed Mar 01 00:00:00 UTC 2023
Spatiotemporal Characterization of Human Early Intervertebral Disc Formation at Single-Cell Resolution.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhou T,Chen Y,Liao Z,Zhang L,Su D,Li Z,Yang X,Ke X,Liu H,Chen Y,Weng R,Shen H,Xu C,Wan Y,Xu R,Su P
Mon May 01 00:00:00 UTC 2023
LC-MS Analysis of Ginsenosides in Different Parts of Panax quinquefolius and Their Potential for Coronary Disease Improvement.
Planta medica
Zhang X,Kong C,Wang X,Hou H,Yu H,Wang L,Li P,Li X,Zhang Y,Han L,Liu K
Thu Jun 01 00:00:00 UTC 2023
Tumor-derived Immunoglobulin-like transcript 4 facilitates angiogenesis of colorectal cancer.
American journal of cancer research
Liu J,Zhang F,He J,Wang S,Wang L,Li J,Shi W,Han Y,Gao A,Sun Y
Sat Mar 11 00:00:00 UTC 2023
IGFBP2 derived from PO-MSCs promote epithelial barrier destruction by activating FAK signaling in nasal polyps.
iScience
You B,Zhang T,Zhang W,Pei Y,Huang D,Lei Y,Zhang S,Qiu C,Zhang J,Gu Z,Cheng L,Chen J
Fri Mar 17 00:00:00 UTC 2023
Gelatin methacryloyl (GelMA) loaded with concentrated hypoxic pretreated adipose-derived mesenchymal stem cells(ADSCs) conditioned medium promotes wound healing and vascular regeneration in aged skin.
Biomaterials research
Li S,Sun J,Yang J,Yang Y,Ding H,Yu B,Ma K,Chen M
Mon Feb 13 00:00:00 UTC 2023
Chitosan oligosaccharide improves ovarian granulosa cells inflammation and oxidative stress in patients with polycystic ovary syndrome.
Frontiers in immunology
Xie Q,Hong W,Li Y,Ling S,Zhou Z,Dai Y,Wu W,Weng R,Zhong Z,Tan J,Zheng Y
Tue Mar 21 00:00:00 UTC 2023
miR-181b promotes angiogenesis and neurological function recovery after ischemic stroke.
Neural regeneration research
Xue LX,Shu LY,Wang HM,Lu KL,Huang LG,Xiang JY,Geng Z,Zhao YW,Chen H
Fri Sep 01 00:00:00 UTC 2023
Network Pharmacology-Based Identification of Key Targets of Ziyin Mingmu Pills Acting on Age-Related Macular Degeneration.
Evidence-based complementary and alternative medicine : eCAM
Yang Y,Wang Y,Liu X,Deng Y,Lu J,Jiang F,Nie F,Peng J,Peng Q,Qin Y
Tue Feb 14 00:00:00 UTC 2023
Antiangiogenic potential of Lepista nuda extract suppressing MAPK/p38 signaling-mediated developmental angiogenesis in zebrafish and HUVECs.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Deshmukh D,Hsu YF,Chiu CC,Jadhao M,Hsu SCN,Hu SY,Yang SH,Liu W
Wed Mar 01 00:00:00 UTC 2023
Network and experimental pharmacology on mechanism of Si-Wu-tang improving ovarian function in a mouse model of premature ovarian failure induced by cyclophosphamide.
Journal of ethnopharmacology
Liu X,Song Y,Zhou F,Zhang C,Li F,Hu R,Ma W,Song K,Tang Z,Zhang M
Mon Jan 30 00:00:00 UTC 2023
USP22 upregulates ZEB1-mediated VEGFA transcription in hepatocellular carcinoma.
Cell death & disease
Zeng K,Xie W,Wang C,Wang S,Liu W,Su Y,Lin L,Zou R,Sun G,Zhou B,Wang M,Luan R,Bai Y,Huo Y,Kato S,Zhong X,Zhao Y
Sat Mar 11 00:00:00 UTC 2023
USP22 upregulates ZEB1-mediated VEGFA transcription in hepatocellular carcinoma.
Cell death & disease
Zeng K,Xie W,Wang C,Wang S,Liu W,Su Y,Lin L,Zou R,Sun G,Zhou B,Wang M,Luan R,Bai Y,Huo Y,Kato S,Zhong X,Zhao Y
Sat Mar 11 00:00:00 UTC 2023
FtMt reduces oxidative stress-induced trophoblast cell dysfunction via the HIF-1α/VEGF signaling pathway.
BMC pregnancy and childbirth
Xu X,Ye X,Zhu M,Zhang Q,Li X,Yan J
Wed Mar 01 00:00:00 UTC 2023
Engineered nanovesicles from stromal vascular fraction promote angiogenesis and adipogenesis inside decellularized adipose tissue through encapsulating growth factors.
Scientific reports
Tu J,Zeng Y,An R,Sun J,Wen H
Fri Jan 13 00:00:00 UTC 2023
Anti-inflammatory and antifibrotic effects of CBP/β-catenin inhibitor for hepatocytes: small molecular inhibitor, OP-724 possibly improves liver function.
Medical molecular morphology
Ouchi H,Mizutani Y,Yoshimura K,Sato Y,Kimura K,Haruyama Y,Harada K
Thu Jun 01 00:00:00 UTC 2023
Chitosan Oligosaccharides Alleviate H2O2-stimulated Granulosa Cell Damage via HIF-1α Signaling Pathway.
Oxidative medicine and cellular longevity
Yang Z,Hong W,Zheng K,Feng J,Hu C,Tan J,Zhong Z,Zheng Y
Tue Apr 12 00:00:00 UTC 2022
The TCM Preparation Feilike Mixture for the Treatment of Pneumonia: Network Analysis, Pharmacological Assessment and Silico Simulation.
Frontiers in pharmacology
Peng J,Chen X,Hou M,Yang K,Yang B,Wang P,Du Y,Yu Q,Ren J,Liu J
Published figure using VEGFA monoclonal antibody (Product # 66828-1-IG) in Immunohistochemistry
Sat Mar 19 00:00:00 UTC 2022
Extracorporeal shockwave relieves endothelial injury and dysfunction in steroid-induced osteonecrosis of the femoral head via miR-135b targeting FOXO1: in vitro and in vivo studies.
Aging
Wu X,Wang Y,Fan X,Xu X,Sun W
Fri Jan 07 00:00:00 UTC 2022
LEM domain containing 1 (LEMD1) transcriptionally activated by SRY-related high-mobility-group box 4 (SOX4) accelerates the progression of colon cancer by upregulating phosphatidylinositol 3-kinase (PI3K)/protein kinase B (Akt) signaling pathway.
Bioengineered
Li D,Wang D,Liu H,Jiang X
Fri Apr 01 00:00:00 UTC 2022
NADH-Cytochrome B5 reductase 2 suppresses retinal vascular dysfunction through regulation of vascular endothelial growth factor A in diabetic retinopathy.
Experimental eye research
Chen J,Sun Y,Chen L,Zhou Y
Thu Sep 01 00:00:00 UTC 2022
Membrane progesterone receptor α (mPRα) enhances hypoxia-induced vascular endothelial growth factor secretion and angiogenesis in lung adenocarcinoma through STAT3 signaling.
Journal of translational medicine
Xia Z,Xiao J,Dai Z,Chen Q
Sat Feb 05 00:00:00 UTC 2022
Inhibitory effects of icotinib combined with antiangiogenic drugs in human non-small cell lung cancer xenograft models are better than single target drugs.
Thoracic cancer
Jiang P,Zhang Y,Cui J,Wang X,Li Y
Sat Jan 01 00:00:00 UTC 2022
Naoluo Xintong Decoction in the Treatment of Ischemic Stroke: A Network Analysis of the Mechanism of Action.
Frontiers in pharmacology
Wang N,Chu F,Fei C,Pan L,Wang Y,Chen W,Peng D,Duan X,He L
Sat Jul 16 00:00:00 UTC 2022
FRAT1 promotes the angiogenic properties of human glioblastoma cells via VEGFA.
Molecular medicine reports
Yang B,Liu D,Ren YQ,Sun YQ,Zhang JP,Wang XG,Wu YQ,Wang SL,Guo SH,Guo G
Tue Mar 01 00:00:00 UTC 2022
Molecular and Cellular Response of the Myocardium (H9C2 Cells) Towards Hypoxia and HIF-1α Inhibition.
Frontiers in cardiovascular medicine
Osuru HP,Lavallee M,Thiele RH
Sat Aug 06 00:00:00 UTC 2022
EPC infusion ameliorates acute graft-versus-host disease-related endothelial injury after allogeneic bone marrow transplantation.
Frontiers in immunology
Wang W,Ye Y,Du Y,Xu Z,Yuan K,Wang Y,Adzraku SY,Li Y,Xu K,Qiao J,Ju W,Zeng L
Wed Jan 04 00:00:00 UTC 2023
Circulating Exosomal miR-181b-5p Promoted Cell Senescence and Inhibited Angiogenesis to Impair Diabetic Foot Ulcer via the Nuclear Factor Erythroid 2-Related Factor 2/Heme Oxygenase-1 Pathway.
Frontiers in cardiovascular medicine
Wang S,Shi M,Zhou J,Wang W,Zhang Y,Li Y
Tue May 10 00:00:00 UTC 2022
Irisin Attenuates Pathological Neovascularization in Oxygen-Induced Retinopathy Mice.
Investigative ophthalmology & visual science
Zhang J,Liu Z,Wu H,Chen X,Hu Q,Li X,Luo L,Ye S,Ye J
Wed Jun 01 00:00:00 UTC 2022
KPNA2 promotes angiogenesis by regulating STAT3 phosphorylation.
Journal of translational medicine
Jia Y,Wang Q,Liang M,Huang K
Wed Dec 28 00:00:00 UTC 2022
HIF-2α regulates proliferation, invasion, and metastasis of hepatocellular carcinoma cells via VEGF/Notch1 signaling axis after insufficient radiofrequency ablation.
Frontiers in oncology
Yang Y,Chen W,Mai W,Gao Y
Tue Oct 11 00:00:00 UTC 2022
HIF-2α regulates proliferation, invasion, and metastasis of hepatocellular carcinoma cells via VEGF/Notch1 signaling axis after insufficient radiofrequency ablation.
Frontiers in oncology
Yang Y,Chen W,Mai W,Gao Y
Tue Oct 11 00:00:00 UTC 2022
Endoglin Modulates TGFβR2 Induced VEGF and Proinflammatory Cytokine Axis Mediated Angiogenesis in Prolonged DEHP-Exposed Breast Cancer Cells.
Biomedicines
Jadhao M,Chen CL,Liu W,Deshmukh D,Liao WT,Chen JY,Urade R,Tsai EM,Hsu SK,Wang LF,Chiu CC
Thu Feb 10 00:00:00 UTC 2022
Estradiol mediates the interaction of LINC01541 and miR-429 to promote angiogenesis of G1/G2 endometrioid adenocarcinoma in-vitro: A pilot study.
Frontiers in oncology
Qiao D,Qin X,Yang H,Liu X,Liu L,Liu S,Jia Z
Tue Aug 23 00:00:00 UTC 2022
Preeclampsia associated changes in volume density of fetoplacental vessels in Chinese women and mouse model of preeclampsia.
Placenta
Shen X,Wang C,Yue X,Wang Q,Xie L,Huang Z,Huang X,Li J,Xu Y,Chen L,Lye S,Wei Y,Wang Z
Fri Apr 01 00:00:00 UTC 2022
Ascorbic acid 2-glucoside preconditioning enhances the ability of bone marrow mesenchymal stem cells in promoting wound healing.
Stem cell research & therapy
Yi Y,Wu M,Zhou X,Xiong M,Tan Y,Yu H,Liu Z,Wu Y,Zhang Q
Mon Mar 21 00:00:00 UTC 2022
Ascorbic acid 2-glucoside preconditioning enhances the ability of bone marrow mesenchymal stem cells in promoting wound healing.
Stem cell research & therapy
Yi Y,Wu M,Zhou X,Xiong M,Tan Y,Yu H,Liu Z,Wu Y,Zhang Q
Mon Mar 21 00:00:00 UTC 2022
Effects of nanoparticle-mediated Co-delivery of bFGF and VEGFA genes to deep burn wounds: An in vivo study.
Colloids and surfaces. B, Biointerfaces
Zhang Q,Zhang C,Feng L,Wang L,Qi J,Dong Y,Zhou YL,Hu K,Zhang Y
Sat Jan 01 00:00:00 UTC 2022
Effects of nanoparticle-mediated Co-delivery of bFGF and VEGFA genes to deep burn wounds: An in vivo study.
Colloids and surfaces. B, Biointerfaces
Zhang Q,Zhang C,Feng L,Wang L,Qi J,Dong Y,Zhou YL,Hu K,Zhang Y
Sat Jan 01 00:00:00 UTC 2022
A photo-triggering double cross-linked adhesive, antibacterial, and biocompatible hydrogel for wound healing.
iScience
Hu H,Zhai X,Li W,Ji S,Dong W,Chen W,Wei W,Lu Z
Fri Jul 15 00:00:00 UTC 2022
EP300 promotes renal tubular epithelial cell fibrosis by increasing HIF2α expression in diabetic nephropathy.
Cellular signalling
Gong Y,Dou Y,Wang L,Wang X,Zhao Z
Sat Oct 01 00:00:00 UTC 2022
Long non-coding RNA NEAT1 promotes angiogenesis in hepatoma carcinoma via the miR-125a-5p/VEGF pathway.
Open life sciences
Guo J,Yuan Q,Fang Y,Liao J,Zhang Z
Tue Oct 11 00:00:00 UTC 2022
Effect of hydroxyapatite bioceramics on the growth of osteoblasts and HIF-α/VEGF signal axis in partial hypoxia environment in vitro.
Technology and health care : official journal of the European Society for Engineering and Medicine
Liu Y,Gan Z,Hu F
Thu Mar 10 00:00:00 UTC 2022
Compound Tongluo Decoction inhibits endoplasmic reticulum stress-induced ferroptosis and promoted angiogenesis by activating the Sonic Hedgehog pathway in cerebral infarction.
Journal of ethnopharmacology
Hui Z,Wang S,Li J,Wang J,Zhang Z
Sun Jan 30 00:00:00 UTC 2022
Glycometabolic reprogramming-mediated proangiogenic phenotype enhancement of cancer-associated fibroblasts in oral squamous cell carcinoma: role of PGC-1α/PFKFB3 axis.
British journal of cancer
Li X,Jiang E,Zhao H,Chen Y,Xu Y,Feng C,Li J,Shang Z
Mon Aug 01 00:00:00 UTC 2022
Gastrin exerts a protective effect against myocardial infarction via promoting angiogenesis.
Molecular medicine (Cambridge, Mass.)
Fu J,Tang Y,Zhang Z,Tong L,Yue R,Cai L
Thu Aug 19 00:00:00 UTC 2021
VEGF/Src signaling mediated pleural barrier damage and increased permeability contributes to subpleural pulmonary fibrosis.
American journal of physiology. Lung cellular and molecular physiology
Lu YZ,Liang LM,Cheng PP,Xiong L,Wang M,Song LJ,Yu F,He XL,Xiong L,Wang XR,Xin JB,Ye H,Ma WL
Tue Jun 01 00:00:00 UTC 2021
LRIG3 Suppresses Angiogenesis by Regulating the PI3K/AKT/VEGFA Signaling Pathway in Glioma.
Frontiers in oncology
Peng C,Chen H,Li Y,Yang H,Qin P,Ma B,Duan Q,Wang B,Mao F,Guo D
Published figure using VEGFA monoclonal antibody (Product # 66828-1-IG) in Western Blot
Tue Mar 16 00:00:00 UTC 2021
Kochia scoparia Saponin Momordin Ic Modulates HaCaT Cell Proliferation and Apoptosis via the Wnt/β-Catenin Pathway.
Evidence-based complementary and alternative medicine : eCAM
Luo M,Zeng B,Wang H,Yang Z,Peng Y,Zhang Y,Wang C
Sat Jul 31 00:00:00 UTC 2021
Identification of glycolysis related pathways in pancreatic adenocarcinoma and liver hepatocellular carcinoma based on TCGA and GEO datasets.
Cancer cell international
Li J,Zhu C,Yue P,Zheng T,Li Y,Wang B,Meng X,Zhang Y
Fri Feb 19 00:00:00 UTC 2021
The mechanism and candidate compounds of aged citrus peel (chenpi) preventing chronic obstructive pulmonary disease and its progression to lung cancer.
Food & nutrition research
Zhou L,Gu W,Kui F,Gao F,Niu Y,Li W,Zhang Y,Guo L,Wang J,Guo Z,Du G
Sun Apr 24 00:00:00 UTC 2022
The mechanism and candidate compounds of aged citrus peel (chenpi) preventing chronic obstructive pulmonary disease and its progression to lung cancer.
Food & nutrition research
Zhou L,Gu W,Kui F,Gao F,Niu Y,Li W,Zhang Y,Guo L,Wang J,Guo Z,Du G
Sun Apr 24 00:00:00 UTC 2022
Anemoside A3 activates TLR4-dependent M1-phenotype macrophage polarization to represses breast tumor growth and angiogenesis.
Toxicology and applied pharmacology
Yin L,Fan Z,Liu P,Chen L,Guan Z,Liu Y,Luo Y
Wed Dec 01 00:00:00 UTC 2021
Nox2 impairs VEGF-A-induced angiogenesis in placenta via mitochondrial ROS-STAT3 pathway.
Redox biology
Hu C,Wu Z,Huang Z,Hao X,Wang S,Deng J,Yin Y,Tan C
Wed Sep 01 00:00:00 UTC 2021
Deletion of p16 accelerates fracture healing in geriatric mice.
American journal of translational research
Ding Q,Liu H,Liu L,Ma C,Qin H,Wei Y,Ren Y
Thu Nov 18 00:00:00 UTC 2021
Deletion of p16 accelerates fracture healing in geriatric mice.
American journal of translational research
Ding Q,Liu H,Liu L,Ma C,Qin H,Wei Y,Ren Y
Thu Nov 18 00:00:00 UTC 2021
The Protective Role of Yin-Yang 1 in Cardiac Injury and Remodeling After Myocardial Infarction.
Journal of the American Heart Association
Huang Y,Li L,Chen H,Liao Q,Yang X,Yang D,Xia X,Wang H,Wang WE,Chen L,Zeng C
Tue Nov 02 00:00:00 UTC 2021
DNMT1-induced miR-378a-3p silencing promotes angiogenesis via the NF-κB signaling pathway by targeting TRAF1 in hepatocellular carcinoma.
Journal of experimental & clinical cancer research : CR
Zhu B,Chen JJ,Feng Y,Yang JL,Huang H,Chung WY,Hu YL,Xue WJ
Mon Nov 08 00:00:00 UTC 2021
Curcumin promotes the survival of ischemic random skin flaps via autophagy.
American journal of translational research
Tu Z,Jiang X,Li Y,Yang S,Lin D,Shi Y,Mao C,Zhang X,Lin C
Tue Apr 13 00:00:00 UTC 2021
Curcumin promotes the survival of ischemic random skin flaps via autophagy.
American journal of translational research
Tu Z,Jiang X,Li Y,Yang S,Lin D,Shi Y,Mao C,Zhang X,Lin C
Tue Apr 13 00:00:00 UTC 2021
The Inhibition of B7H3 by 2-HG Accumulation Is Associated With Downregulation of VEGFA in IDH Mutated Gliomas.
Frontiers in cell and developmental biology
Zhang M,Zhang H,Fu M,Zhang J,Zhang C,Lv Y,Fan F,Zhang J,Xu H,Ye D,Yang H,Hua W,Mao Y
Sat Jun 05 00:00:00 UTC 2021
The Inhibition of B7H3 by 2-HG Accumulation Is Associated With Downregulation of VEGFA in IDH Mutated Gliomas.
Frontiers in cell and developmental biology
Zhang M,Zhang H,Fu M,Zhang J,Zhang C,Lv Y,Fan F,Zhang J,Xu H,Ye D,Yang H,Hua W,Mao Y
Sat Jun 05 00:00:00 UTC 2021
Tubeimoside I promotes angiogenesis via activation of eNOS-VEGF signaling pathway.
Journal of ethnopharmacology
Yang X,Li X,Luo M,Guo Y,Li C,Lv D,Cheng Z,Huang L,Shang FF,Huang B,Shen J,Luo S,Yan J
Mon Mar 01 00:00:00 UTC 2021
Ginsenoside CK Inhibits Hypoxia-Induced Epithelial-Mesenchymal Transformation through the HIF-1α/NF-κB Feedback Pathway in Hepatocellular Carcinoma.
Foods (Basel, Switzerland)
Zhang J,Ma X,Fan D
Wed May 26 00:00:00 UTC 2021
Effects of S1PR2 antagonist on blood pressure and angiogenesis imbalance in preeclampsia rats.
Molecular medicine reports
Zhang T,Guo D,Zheng W,Dai Q
Tue Jun 01 00:00:00 UTC 2021
Upregulation of microfibrillar-associated protein 2 is closely associated with tumor angiogenesis and poor prognosis in hepatocellular carcinoma.
Oncology letters
Zhang N,Shao F,Jia W
Fri Oct 01 00:00:00 UTC 2021
ARHGAP20 Expression Inhibited HCC Progression by Regulating the PI3K-AKT Signaling Pathway.
Journal of hepatocellular carcinoma
Liu G,Li J,Zhang CY,Huang DY,Xu JW
Fri Apr 22 00:00:00 UTC 2022
Ruyiping extract reduces lung metastasis in triple negative breast cancer by regulating macrophage polarization.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Yang R,Xie Y,Li Q,Ye Y,Shi Y,Zhao X,Wu C,Xu Y,Wang R,Zhang Y,Yang X,Han X,Liu S
Wed Sep 01 00:00:00 UTC 2021
Effect of Babao Dan on angiogenesis of gastric cancer in vitro by regulating VEGFA/VEGFR2 signaling pathway.
Translational cancer research
Guan JH,Cao ZY,Guan B,Wei LH,Peng J,Chen YQ,Sferra TJ,Sankararaman S,Zhan ZX,Lin JM
Mon Feb 01 00:00:00 UTC 2021
Conditioned medium from induced pluripotent stem cell-derived mesenchymal stem cells accelerates cutaneous wound healing through enhanced angiogenesis.
Stem cell research & therapy
Liang X,Lin F,Ding Y,Zhang Y,Li M,Zhou X,Meng Q,Ma X,Wei L,Fan H,Liu Z
Thu May 20 00:00:00 UTC 2021
Exosomal circRNAs contribute to intestinal development via the VEGF signalling pathway in human term and preterm colostrum.
Aging
Zhou Y,Yu Z,Wang X,Chen W,Liu Y,Zhang Y,Yin J,Han S
Sun Apr 04 00:00:00 UTC 2021
Protective function of exosomes from adipose tissue-derived mesenchymal stem cells in acute kidney injury through SIRT1 pathway.
Life sciences
Gao F,Zuo B,Wang Y,Li S,Yang J,Sun D
Sat Aug 15 00:00:00 UTC 2020
Hirudin Reduces the Expression of Markers of the Extracellular Matrix in Renal Tubular Epithelial Cells in a Rat Model of Diabetic Kidney Disease Through the Hypoxia-Inducible Factor-1α (HIF-1α)/Vascular Endothelial Growth Factor (VEGF) Signaling Pathway.
Medical science monitor : international medical journal of experimental and clinical research
Pang X,Zhang Y,Shi X,Peng Z,Xing Y,Jiarui H
Sat May 30 00:00:00 UTC 2020
Effect of the Soluble Factors Released by Dental Apical Papilla-Derived Stem Cells on the Osteo/Odontogenic, Angiogenic, and Neurogenic Differentiation of Dental Pulp Cells.
Stem cells and development
Yu S,Zhao Y,Fang TJ,Ge L
Mon Jun 15 00:00:00 UTC 2020
MicroRNA‑21 contributes to renal cell carcinoma cell invasiveness and angiogenesis via the PDCD4/c‑Jun (AP‑1) signalling pathway.
International journal of oncology
Fan B,Jin Y,Zhang H,Zhao R,Sun M,Sun M,Yuan X,Wang W,Wang X,Chen Z,Liu W,Yu N,Wang Q,Liu T,Li X
Wed Jan 01 00:00:00 UTC 2020
MiR-103a-3p promotes tumour glycolysis in colorectal cancer via hippo/YAP1/HIF1A axis.
Journal of experimental & clinical cancer research : CR
Sun Z,Zhang Q,Yuan W,Li X,Chen C,Guo Y,Shao B,Dang Q,Zhou Q,Wang Q,Wang G,Liu J,Kan Q
Fri Nov 20 00:00:00 UTC 2020
circ_2858 Helps Blood-Brain Barrier Disruption by Increasing VEGFA via Sponging miR-93-5p during Escherichia coli Meningitis.
Molecular therapy. Nucleic acids
Yang R,Chen J,Xu B,Yang B,Fu J,Xiao S,Tan C,Chen H,Wang X
Fri Dec 04 00:00:00 UTC 2020
miR-302e Suppresses Glioma Progression by Targeting VEGFA.
Cancer management and research
Xie Y,Liu X,Hu T,Wang W
Mon Apr 18 00:00:00 UTC 2022
TMSB10 acts as a biomarker and promotes progression of clear cell renal cell carcinoma.
International journal of oncology
Pan Q,Cheng G,Liu Y,Xu T,Zhang H,Li B
Fri May 01 00:00:00 UTC 2020
Proliferation inhibition and apoptosis promotion by dual silencing of VEGF and Survivin in human osteosarcoma.
Acta biochimica et biophysica Sinica
Gu J,Ji Z,Li D,Dong Q
Tue Jan 01 00:00:00 UTC 2019
Proliferation inhibition and apoptosis promotion by dual silencing of VEGF and Survivin in human osteosarcoma.
Acta biochimica et biophysica Sinica
Gu J,Ji Z,Li D,Dong Q
Tue Jan 01 00:00:00 UTC 2019
Proliferation inhibition and apoptosis promotion by dual silencing of VEGF and Survivin in human osteosarcoma.
Acta biochimica et biophysica Sinica
Gu J,Ji Z,Li D,Dong Q
Tue Jan 01 00:00:00 UTC 2019
Proliferation inhibition and apoptosis promotion by dual silencing of VEGF and Survivin in human osteosarcoma.
Acta biochimica et biophysica Sinica
Gu J,Ji Z,Li D,Dong Q
Tue Jan 01 00:00:00 UTC 2019
Dimethyl fumarate suppresses hepatocellular carcinoma progression via activating SOCS3/JAK1/STAT3 signaling pathway.
American journal of translational research
Liu H,Feng XD,Yang B,Tong RL,Lu YJ,Chen DY,Zhou L,Xie HY,Zheng SS,Wu J
Thu Oct 01 00:00:00 UTC 2020
Targeting Angiogenesis by Blocking the ATM-SerRS-VEGFA Pathway for UV-Induced Skin Photodamage and Melanoma Growth.
Cancers
Song Y,Lu H,Wang Q,Xiang R
Fri Nov 22 00:00:00 UTC 2019
Targeting Angiogenesis by Blocking the ATM-SerRS-VEGFA Pathway for UV-Induced Skin Photodamage and Melanoma Growth.
Cancers
Song Y,Lu H,Wang Q,Xiang R
Fri Nov 22 00:00:00 UTC 2019