Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: GDEGAEAPVE
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA and glycine transporter; GVAT; hVIAAT; rat UNC-47 homolog; rGVAT; RUNC-47; solute carrier family 32 (GABA vesicular transporter), member 1; Solute carrier family 32 member 1; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter
Gene Aliases: SLC32A1; VGAT; VIAAT
UniProt ID: (Human) Q9H598, (Rat) O35458
Entrez Gene ID: (Human) 140679, (Rat) 83612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support