Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METPLDVLSR AASLVHADDE KREAALRGEP RMQTLPVASA LSSHRTGPPP ISPSKRKFSM EPGDEDLDCD NDHVSKMSRI FNPHLNKTAN GDCRRDPRER SRSPIERAVA PTMSLHGSHL YTSLPSLGLE QPLALTKNSL DASRPAGLSP TLTPGERQQN RPSVITCASA GARNCNLSHC PIAHSGCAAP GPASYRRPPS AATTCDPVVE EHFRRSLGKN YKEPEPAPNS VSITGSVDDH FAKALGDTWL QIKAAKDGAS SSPESASRRG QPASPSAHMV SHSHSPSVVS
Transcription cofactor vestigial-like protein 4 (VGLL4) acts as a specific coactivator for the mammalian TEFs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transcription cofactor vestigial-like protein 4; vestigial like 4; Vestigial-like 4; vestigial-like family member 4; Vgl-4
Gene Aliases: KIAA0121; VGL-4; VGLL4
UniProt ID: (Human) Q14135
Entrez Gene ID: (Human) 9686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support