Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KHKTREEWQY VFLIASLVHY GGVIFYGVFA SGEKQPWAEP EEMSEEKCGF VGHDQLAGSD DSEMEDEAEP PGAPPAPPPS YGATHSTFQP PRPPPPVRDY; Positive Samples: SH-SY5Y; Cellular Location: Cell junction, Cytoplasmic vesicle, Membrane, Multi-pass membrane protein, secretory vesicle, synapse, synaptic vesicle membrane, synaptosome
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGluT1
Gene Aliases: 2900052E22Rik; AI851913; BNPI; SLC17A7; VGLUT1
UniProt ID: (Human) Q9P2U7, (Mouse) Q3TXX4
Entrez Gene ID: (Human) 57030, (Mouse) 72961
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support