Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
1 µg/mL of MA5-27594 was sufficient for detection of VGLUT3 in 20 µg of CV-1 fibroblast cells (lysate) transfected with VGLUT3 by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 65kDa.
This antibody was formerly sold as clone S34-34.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT-3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; Solute carrier family 17 member 8; Vesicular glutamate transporter 3; vesicular glutamate transporter-3; VGLUT 3; VGluT3
Gene Aliases: BC042593; DFNA25; SLC17A8; VGLUT3
UniProt ID: (Human) Q8NDX2, (Mouse) Q8BFU8, (Rat) Q7TSF2
Entrez Gene ID: (Human) 246213, (Mouse) 216227, (Rat) 266767
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support