Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: BxPC-3, U-87MG, SH-SY5Y, Mouse spleen, Mouse brain, Rat brain. The target is usually found in the following locations: Secreted.
Immunogen sequence: MDTRNKAQLL VLLTLLSVLF SQTSAWPLYR APSALRLGDR IPFEGANEPD QVSLKEDIDM LQNALAENDT PYYDVSRNAR HADGVFTSDF SKLLGQLSAK KYLESLMGKR VSNISEDPVP VKRHSDAVFT DNYTRLRKQM AVKKYLNSIL NGKRSSEGES PDFPEELEK
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC13587; PACAP-R-2; PHI, peptide histidine isoleucine; prepro-VIP; vasoactive intestinal polypeptide; VIP peptides
Gene Aliases: PHM27; VIP; vip/phi27
UniProt ID: (Human) P01282, (Rat) P01283, (Mouse) P32648
Entrez Gene ID: (Human) 7432, (Rat) 117064, (Mouse) 22353
Molecular Function:
neuropeptide
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support