Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Vitamin K epoxide reductase complex subunit 1 (VKORC1) is the enzyme that is responsible for reducing vitamin K 2,3-epoxide to the enzymatically activated form which is essential for blood clotting. This enzymatically activated form of vitamin K is a reduced form required for the carboxylation of glutamic acid residues in some blood-clotting proteins. Fatal bleeding can be caused by vitamin K deficiency and by the vitamin K antagonist warfarin, and it is VKORC1 that is sensitive to warfarin. In humans, mutations in this gene can be associated with deficiencies in vitamin-K-dependent clotting factors and, in humans and rats, with warfarin resistance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMAGE3455200; phylloquinone epoxide reductase; vitamin K dependent clotting factors deficiency 2; Vitamin K epoxide reductase complex subunit 1; vitamin K epoxide reductase complex, subunit 1; Vitamin K1 2,3-epoxide reductase subunit 1; vitamin K1 epoxide reductase (warfarin-sensitive); Warfarin resistance
Gene Aliases: D7Wsu86e; EDTP308; MST134; MST576; MSTP134; MSTP576; UNQ308/PRO351; VKCFD2; VKOR; VKORC1
UniProt ID: (Human) Q9BQB6, (Rat) Q6TEK4, (Mouse) Q9CRC0
Entrez Gene ID: (Human) 79001, (Rat) 309004, (Mouse) 27973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support