Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Zebrafish, X. tropicalis, Chicken, Bovine and Orangutan
Required for the assembly of the V0 complex of the vacuolar ATPase (V-ATPase) in the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myopathy with excessive autophagy protein; Vacuolar ATPase assembly integral membrane protein VMA21
Gene Aliases: 2610030H06Rik; AI840175; MEAX; VMA21; XMEA
UniProt ID: (Human) Q3ZAQ7, (Mouse) Q78T54
Entrez Gene ID: (Human) 203547, (Mouse) 67048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support