Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: VMAT2.
VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC120477; MGC120478; MGC26538; Monoamine transporter; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); Solute carrier family 18 member 2; solute carrier family 18, member 2; Synaptic vesicular amine transporter; VAT2; Vesicular amine transporter 2; Vesicular monoamine transporter 2
Gene Aliases: 1110037L13Rik; 9330105E13; MNAT; Slc18a2; Svat; VMAT-2; VMAT2
UniProt ID: (Mouse) Q8BRU6, (Rat) Q01827
Entrez Gene ID: (Mouse) 214084, (Rat) 25549
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support