Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Putative pheromone receptor. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor GPCR25; hGPCR25; pheromone receptor; V1R-like 2; V1r-like receptor 2; Vomeronasal type-1 receptor 2
Gene Aliases: V1RL2; VN1R2
UniProt ID: (Human) Q8NFZ6
Entrez Gene ID: (Human) 317701
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support